Skip to Content
Merck
All Photos(1)

Documents

G4376

Sigma-Aldrich

L-Glutathione oxidized

≥98% (HPLC)

Synonym(s):

GSSG, Glutathiol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H32N6O12S2
CAS Number:
Molecular Weight:
612.63
Beilstein:
1718700
EC Number:
MDL number:
UNSPSC Code:
12161501
PubChem Substance ID:
NACRES:
NA.21

Assay

≥98% (HPLC)

form

powder

solubility

H2O: soluble

storage temp.

2-8°C

SMILES string

N[C@@H](CCC(=O)N[C@@H](CSSC[C@H](NC(=O)CC[C@H](N)C(O)=O)C(=O)NCC(O)=O)C(=O)NCC(O)=O)C(O)=O

InChI

1S/C20H32N6O12S2/c21-9(19(35)36)1-3-13(27)25-11(17(33)23-5-15(29)30)7-39-40-8-12(18(34)24-6-16(31)32)26-14(28)4-2-10(22)20(37)38/h9-12H,1-8,21-22H2,(H,23,33)(H,24,34)(H,25,27)(H,26,28)(H,29,30)(H,31,32)(H,35,36)(H,37,38)/t9-,10-,11-,12-/m0/s1

InChI key

YPZRWBKMTBYPTK-BJDJZHNGSA-N

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Glu-Cys-Gly, Glu-Cys-Gly [Disulfide bridge: 2a-2b]

General description

Our SAFC® portfolio of high-quality raw materials for use in biopharmaceutical processing withstands strict quality control procedures plus the documentation and expertise to help our customers meet requirements as defined by the M-Clarity Program.

M-Clarity Program

Our comprehensive portfolio of downstream process chemicals not only provides biopharmaceutical manufacturers with high-quality raw materials for production of classical and novel therapies, but also helps them get to market faster and simplify regulatory challenges.

Ranging from non-GMP grades for low-risk application, to IPEC-PQG GMP for higher-risk applications, we have products covering all your manufacturing needs.

Legal Information

SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N O Kaakoush et al.
Antimicrobial agents and chemotherapy, 53(5), 1884-1891 (2009-02-19)
Metronidazole resistance in Helicobacter pylori has been attributed to mutations in rdxA or frxA. Insufficient data correlating RdxA and/or FrxA with the resistant phenotype, and the emergence of resistant strains with no mutations in either rdxA or frxA, indicated that
Roman Kireev et al.
European journal of pharmacology, 701(1-3), 185-193 (2012-12-12)
Fatty livers occur in up to 20% of potential liver donors and increase cellular injury during the ischemia/reperfusion phase, so any intervention that could enable a better outcome of grafts for liver transplantation would be very useful. The effect of
Patrick M McNutt et al.
Science translational medicine, 13(575) (2021-01-08)
Botulism is caused by a potent neurotoxin that blocks neuromuscular transmission, resulting in death by asphyxiation. Currently, the therapeutic options are limited and there is no antidote. Here, we harness the structural and trafficking properties of an atoxic derivative of
Ella Doron-Mandel et al.
The EMBO journal, 40(20), e107158-e107158 (2021-09-14)
Nucleolin is a multifunctional RNA Binding Protein (RBP) with diverse subcellular localizations, including the nucleolus in all eukaryotic cells, the plasma membrane in tumor cells, and the axon in neurons. Here we show that the glycine arginine rich (GAR) domain
Raisa Turja et al.
Environmental science and pollution research international, 27(13), 15498-15514 (2020-02-23)
In the brackish water Baltic Sea, oil pollution is an ever-present and significant environmental threat mainly due to the continuously increasing volume of oil transport in the area. In this study, effects of exposure to crude oil on two common

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service