Skip to Content
Merck
All Photos(3)

Key Documents

M2773

Sigma-Aldrich

Magnesium sulfate heptahydrate

BioReagent, for molecular biology, suitable for plant cell culture, ≥99.0%

Synonym(s):

Epsom salts

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MgSO4 · 7H2O
CAS Number:
Molecular Weight:
246.47
EC Number:
MDL number:
UNSPSC Code:
12352302
eCl@ss:
38030409
PubChem Substance ID:
NACRES:
NA.31
grade:
Molecular Biology
for molecular biology

grade

Molecular Biology
for molecular biology

vapor density

<0.01 (vs air)

vapor pressure

<0.1 mmHg ( 20 °C)

product line

BioReagent

Assay

≥99.0%

technique(s)

cell culture | plant: suitable

anion traces

chloride (Cl-): ≤5 ppm

cation traces

K: ≤50 ppm
Na: ≤50 ppm
heavy metals (as Pb): ≤5 ppm

foreign activity

DNase, RNase and protease, none detected

SMILES string

O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O

InChI

1S/Mg.H2O4S.7H2O/c;1-5(2,3)4;;;;;;;/h;(H2,1,2,3,4);7*1H2/q+2;;;;;;;;/p-2

InChI key

WRUGWIBCXHJTDG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Magnesium sulfate have anti-convulsant, anti-inflammatory and vasodilatory properties. Magnesium sulfate relaxes smooth muscles. In acute asthma patient, both intravenous and nebulization of magnesium sulfate as an additive improves pulmonary function. Magnesium sulfate reduces preterm labor in high risk pregnant women. Magnesium sulfate heptahydrate serves as a source of Mg2+ ions in plant tissue culture medium, and protoplast culture medium.

Application

Magnesium sulfate heptahydrate has been used:
  • in the preparation of Scott′s tap water substitute as a bluing reagent for nuclear staining
  • as a component of stabilization solution for the isolation of pituitary cells for matrix-assisted laser desorption/ionization mass spectroscopy (MALDI-MS)
  • as an additive in the preparation of artificial sea water for the maintenance of Aplysia californica and in modified Gey′s balanced salt solution

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characterizing peptides in individual mammalian cells using mass spectrometry
Rubakhin SS and Sweedler JV
Nature Protocols, 2(8), 1987-1987 (2007)
Qualitative and quantitative metabolomic investigation of single neurons by capillary electrophoresis electrospray ionization mass spectrometry
Nemes, Peter and Rubakhin, Stanislav S and Aerts, Jordan T and Sweedler, Jonathan V
Nature Protocols, 8(4), 783-783 (2013)
Protoplast isolation, culture and somatic hybridization
Plant Tissue Culture: Theory and Techniques (2016)
Embedding, serial sectioning and staining of zebrafish embryos using JB-4 resin
Sullivan-Brown J, et al.
Nature Protocols, 6(1), 46-46 (2011)
Getting started with tissue culture-Media preparation, sterile technique and laboratory equipment
Plant Tissue Culture, Development, and Biotechnology, 15-15 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service