Skip to Content
Merck
All Photos(2)

Key Documents

M7899

Sigma-Aldrich

Manganese(II) sulfate monohydrate

BioReagent, suitable for plant cell culture, suitable for cell culture

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MnSO4 · H2O
CAS Number:
Molecular Weight:
169.02
EC Number:
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.72
form:
powder

product line

BioReagent

Quality Level

form

powder

technique(s)

cell culture | mammalian: suitable
cell culture | plant: suitable

application(s)

agriculture

SMILES string

O.[Mn++].[O-]S([O-])(=O)=O

InChI

1S/Mn.H2O4S.H2O/c;1-5(2,3)4;/h;(H2,1,2,3,4);1H2/q+2;;/p-2

InChI key

ISPYRSDWRDQNSW-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Manganese sulfate is present in nature as numerous mineral forms such as, jokokuite, pentahydrite, szmikite, and mallardite. Manganese is found in various superoxide dismutases.

Application

Manganese (II) sulfate monohydrate has been used as a micronutrient for preparation of nutritive medium for growth of sugarcane plantlets. It has also been used as a trace element in the preparation of N6 complete nutrient liquid medium for growing mycelia of H. cylindrosporum.
Manganese(II) sulfate monohydrate is plant cell culture tested (0.6 g/ml) and is appropriate for use in plant cell culture experiments. Manganese (II) sulfate is a component of several plant cell culture media including: Chu′s N6; DKW/Juglans; Murashige and Skoog; and Gamborg′s B-5 media.

Biochem/physiol Actions

Manganese sulfate has various industrial applications such as dyeing, porcelain glazing, and the manufacture of fertilizers and boiling oils. MnSO4 is used as a source of manganese ion in biological research, such as in culturing of Bacillus licheniformis and the induction of chromosomal abnormalities in plants. MnSO4 has been utilized to investigate the enzyme dependent glycosylation of endogenous glycoproteins in human skeletal muscle.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - STOT RE 2 Inhalation

Target Organs

Brain

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Method for Radioactive labelling of Hebeloma cylindrosporum to study plant-fungus interactions
Becquer A, et al.
Bio-protocol, 7(20), e2576-e2576 (2017)
Bioassay to Study the Attachment of Xanthomonas albilineans on Sugarcane Leaves
Mensi I, et al.
Bio-protocol, 7(2) (2017)
Jee-Hwan Oh et al.
Cell host & microbe, 25(2), 273-284 (2019-01-20)
The mammalian intestinal tract contains a complex microbial ecosystem with many lysogens, which are bacteria containing dormant phages (prophages) inserted within their genomes. Approximately half of intestinal viruses are derived from lysogens, suggesting that these bacteria encounter triggers that promote
Imène Mensi et al.
Bio-protocol, 7(2), e2111-e2111 (2017-01-20)
Sugarcane (interspecific hybrids of Saccharum species) is an economically important crop that provides 70% of raw table sugar production worldwide and contributes, in some countries, to bioethanol and electricity production. Leaf scald, caused by the bacterial plant pathogen Xanthomonas albilineans
Haixing Li et al.
Amino acids, 38(5), 1439-1445 (2009-09-30)
Production of gamma-aminobutyric acid (GABA) was carried out in Erlenmeyer flasks by Lactobacillus brevis NCL912. Traditional methods were first adopted to select the key factors that impact the GABA production to preliminarily determine the suitable concentration ranges of the key

Protocols

Reference guide and preparation guide for antibiotic and antimycotic use in plant tissue culture.

Reference guide and preparation guide for antibiotic and antimycotic use in plant tissue culture.

Reference guide and preparation guide for antibiotic and antimycotic use in plant tissue culture.

Reference guide and preparation guide for antibiotic and antimycotic use in plant tissue culture.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service