Skip to Content
Merck
All Photos(1)

Key Documents

63140

Sigma-Aldrich

Magnesium sulfate heptahydrate

puriss. p.a., ACS reagent, ≥99.0% (KT)

Synonym(s):

Epsom salts

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MgSO4 · 7H2O
CAS Number:
Molecular Weight:
246.47
EC Number:
MDL number:
UNSPSC Code:
12352302
eCl@ss:
38030409
PubChem Substance ID:
NACRES:
NB.24
Assay:
≥99.0% (KT)
form:
powder or crystals

grade

ACS reagent
puriss. p.a.

Quality Level

vapor density

<0.01 (vs air)

vapor pressure

<0.1 mmHg ( 20 °C)

Assay

≥99.0% (KT)

form

powder or crystals

impurities

≤0.005% insoluble matter

pH

5.0-8.2 (25 °C, 50 mg/mL in H2O)

anion traces

chloride (Cl-): ≤5 mg/kg
nitrate (NO3-): ≤20 mg/kg

cation traces

As: ≤0.1 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤1 mg/kg
K: ≤10 mg/kg
Mn: ≤5 mg/kg
NH4+: ≤20 mg/kg
Na: ≤10 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg
heavy metals: ≤5 ppm (by ICP-OES)

SMILES string

O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O

InChI

1S/Mg.H2O4S.7H2O/c;1-5(2,3)4;;;;;;;/h;(H2,1,2,3,4);7*1H2/q+2;;;;;;;;/p-2

InChI key

WRUGWIBCXHJTDG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Magnesium sulfate heptahydrate can be used as a catalyst to synthesize tri/tetra-substituted imidazoles via a one-pot condensation reaction of aromatic aldehydes, benzil, aliphatic and aromatic primary amines, and ammonium acetate. It is also used to synthesize phenazine and quinoxaline derivatives.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

ESR studies of (Cu(H2O)6)2+ in magnesium sulfate heptahydrate.
Radhakrishna S and Bhaskar Rao T.
Journal of Magnetic Resonance, 32(1), 71-81 (1978)
Sulfates on Mars: A systematic Raman spectroscopic study of hydration states of magnesium sulfates.
Wang A, et al.
Geochimica et Cosmochimica Acta, 70(24), 6118-6135 (2006)
Kirsten M Ellegaard et al.
Current biology : CB, 30(13), 2520-2531 (2020-06-13)
Most bacterial species encompass strains with vastly different gene content. Strain diversity in microbial communities is therefore considered to be of functional importance. Yet little is known about the extent to which related microbial communities differ in diversity at this
Petra Marhava et al.
Developmental cell, 52(2), 223-235 (2019-12-24)
Cell polarity is a key feature in the development of multicellular organisms. For instance, asymmetrically localized plasma-membrane-integral PIN-FORMED (PIN) proteins direct transcellular fluxes of the phytohormone auxin that govern plant development. Fine-tuned auxin flux is important for root protophloem sieve
Philippe Jean et al.
Nature communications, 11(1), 3208-3208 (2020-06-27)
Inner hair cells (IHCs) are the primary receptors for hearing. They are housed in the cochlea and convey sound information to the brain via synapses with the auditory nerve. IHCs have been thought to be electrically and metabolically independent from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service