Skip to Content
Merck
All Photos(3)

Key Documents

M2643

Sigma-Aldrich

Magnesium sulfate

BioReagent, suitable for cell culture, suitable for insect cell culture

Synonym(s):

Magnesium Sulphate, Magnesium(II) sulfate, MgSO4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
MgSO4
CAS Number:
Molecular Weight:
120.37
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.71
grade:
reagent
form:
powder

grade

reagent

vapor density

<0.01 (vs air)

vapor pressure

<0.1 mmHg ( 20 °C)

product line

BioReagent

form

powder

technique(s)

cell culture | insect: suitable
cell culture | mammalian: suitable

SMILES string

[Mg++].[O-]S([O-])(=O)=O

InChI

1S/Mg.H2O4S/c;1-5(2,3)4/h;(H2,1,2,3,4)/q+2;/p-2

InChI key

CSNNHWWHGAXBCP-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Magnesium sulphate reduces eclampsia in women. It may inhibit N-methyl-D-aspartate (NMDA) receptor to accomplish the anticonvulsant activity.

Application

Magnesium sulphate has been used:
  • as a component of M63 working medium solution for E. coli microcolonies in gel beads and Synechocystis liquid growth medium
  • as a buffer component for phosphoglycerate mutase assay and ribokinase activity assay
  • in ice-cold assay buffer for plasma membrane cholesterol measurement assay in immortalized bone marrow-derived macrophages (iBMDMs)

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Deuterated nucleotides as chemical probes of RNA structure: a detailed protocol for the enzymatic synthesis of a complete set of nucleotides specifically deuterated at ribose carbons
Tullius T, et al.
ScienceOpen Research (2015)
Bacterial Microcolonies in Gel Beads for High-throughput Screening
Schaerli Y
Bio-protocol, 8(13) (2018)
Magnesium sulphate and other anticonvulsants for women with pre-eclampsia
Duley L, et al.
Cochrane database of systematic reviews (Online) (2010)
Analysis of Metals in Whole Cells, Thylakoids and Photosynthetic Protein Complexes in Synechocystis sp. PCC6803
Gandini C, et al.
Bio-protocol, 8(12) (2018)
Magnesium sulfate in eclampsia and pre-eclampsia
Lu JF and Nightingale CH
Clinical Pharmacokinetics, 38(4), 305-314 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service