Skip to Content
Merck
All Photos(3)

Documents

F8633

Sigma-Aldrich

Iron(II) sulfate heptahydrate

BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%

Synonym(s):

Ferrous sulfate heptahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FeSO4 · 7H2O
CAS Number:
Molecular Weight:
278.01
EC Number:
MDL number:
UNSPSC Code:
12352207
PubChem Substance ID:
NACRES:
NA.71

vapor pressure

14.6 mmHg ( 25 °C)

product line

BioReagent

Assay

≥99%

shelf life

2 yr

technique(s)

cell culture | insect: suitable
cell culture | mammalian: suitable

density

1.898 g/mL at 25 °C (lit.)

SMILES string

O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O

InChI

1S/Fe.H2O4S.7H2O/c;1-5(2,3)4;;;;;;;/h;(H2,1,2,3,4);7*1H2/q+2;;;;;;;;/p-2

InChI key

SURQXAFEQWPFPV-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Iron(II) sulfate heptahydrate is used in cell culture applications generally bound to transferrin, citrate or other iron transport molecules.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zahidul Alam et al.
Cell reports, 31(13), 107825-107825 (2020-07-02)
Activated macrophages must carefully calibrate their inflammatory responses to balance efficient pathogen control with inflammation-mediated tissue damage, but the molecular underpinnings of this "balancing act" remain unclear. Using genetically engineered mouse models and primary macrophage cultures, we show that Toll-like
Ya-Na Wu et al.
International journal of nanomedicine, 8, 3321-3331 (2013-09-17)
Previously, iron core-gold shell nanoparticles (Fe@Au) have been shown to possess cancer-preferential cytotoxicity in oral and colorectal cancer (CRC) cells. However, CRC cell lines are less sensitive to Fe@Au treatment when compared with oral cancer cell lines. In this research
Oli A Fulas et al.
Journal of neurochemistry, 152(5), 570-584 (2019-12-20)
Local microvascular dysfunction and consequent tissue ischemia/hypoxia contribute to the symptoms of complex regional pain syndrome (CRPS) and peripheral neuropathic pain. As nitric oxide (NO) is a key regulator of microvascular blood flow, compounds that increase it are potentially therapeutic
Jung-Feng Lin et al.
International journal of molecular sciences, 20(21) (2019-10-28)
Chronic renal failure involving hemodialysis results in blood loss during filtration. Iron deficiency and iron deficiency anemia can result. A compensatory increase in iron dosage has many side effects including discomfort. Elemental iron is a highly-pure iron source, which reduces
Xiao-Kui Ma et al.
Carbohydrate polymers, 92(2), 1188-1196 (2013-02-13)
The influence of several components on exopolysaccharide (EPS) production and antioxidative activity (TEAC, Trolox-Equivalent Antioxidant Capacity) as well as their effects on the morphological development and cell viability of Phellinus sp. P0988 was determined. Rutin, FeSO(4), Tween 80 and complex

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service