Skip to Content
Merck
All Photos(1)

Key Documents

E3024

Sigma-Aldrich

Earle′s Balanced Salts

With sodium bicarbonate, without phenol red, liquid, sterile-filtered, suitable for cell culture

Synonym(s):

EBSS

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.75

Quality Level

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

components

phenol red: no
NaHCO3: 2.2 g/L
glucose: 1.0 g/L (Dextro)

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

General description

Earle′s balanced salts solution (EBSS) is a saline solution with physiological pH that stimulates autophagy. It augments Bacillus Calmette-Guérin (BCG)-induced dendritic cells (DCs) by upregulating cluster of differentiation (CD86), human leukocyte antigen-DR isotype (HLA-DR), and interleukin (IL-6).

Application

Earle′s balanced salts has been used to incubate filter-grown monolayers of Caco-2 cells to induce starvation, along with HEPES buffer to detect NADPH oxidase activity in trypsinized cells. It has also been used to treat CEM RES cells and perform amino acid depletion experiment.

Other Notes

Phenol red has been shown to interfere with the growth of some cells at cloning densities. Use this medium when working with stem cells or when growing cells at low densities.

supplement

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jagadeesan Arunkumar et al.
Microbial pathogenesis, 118, 301-309 (2018-04-01)
Herpes simplex virus - 2 (HSV-2) causes lifelong persisting infection in the immunocompromised host and intermittent in healthy individuals with high morbidity in neonatals and also increase the transmission of HIV. Acyclovir is widely used drug to treat HSV-2 infection
Morgan Wong et al.
American journal of physiology. Cell physiology, 316(5), C753-C765 (2019-03-21)
A defective tight junction (TJ) barrier is a key pathogenic factor for inflammatory bowel disease. Previously, we have shown that autophagy, a cell survival mechanism, enhances intestinal epithelial TJ barrier function. Autophagy-related protein-6 (ATG6/beclin 1), a key protein in the
Natthakan Thongon et al.
Cancer & metabolism, 6, 1-1 (2018-03-16)
Inhibitors of nicotinamide phosphoribosyltransferase (NAMPT), the rate-limiting enzyme in NAD+ biosynthesis from nicotinamide, exhibit anticancer effects in preclinical models. However, continuous exposure to NAMPT inhibitors, such as FK866, can induce acquired resistance. We developed FK866-resistant CCRF-CEM (T cell acute lymphoblastic
Léon-Charles Tranchevent et al.
Genome research, 27(6), 1087-1097 (2017-04-20)
Transcriptomic genome-wide analyses demonstrate massive variation of alternative splicing in many physiological and pathological situations. One major challenge is now to establish the biological contribution of alternative splicing variation in physiological- or pathological-associated cellular phenotypes. Toward this end, we developed
Stanislas Morand et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 23(4), 1205-1218 (2008-12-17)
Dual oxidases (Duox1 and Duox2) are plasma membrane-targeted hydrogen peroxide generators that support extracellular hemoperoxidases. Duox activator 2 (Duoxa2), initially described as an endoplasmic reticulum resident protein, functions as a maturation factor needed to deliver active Duox2 to the cell

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service