Skip to Content
Merck
All Photos(2)

Key Documents

B1793

Sigma-Aldrich

Bafilomycin A1

from Streptomyces griseus, ≥90% (HPLC), film, V-ATPase inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H58O9
CAS Number:
Molecular Weight:
622.83
Beilstein:
4730700
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Product Name

Bafilomycin A1 from Streptomyces griseus, ≥90% (HPLC)

Quality Level

Assay

≥90% (HPLC)

form

film

antibiotic activity spectrum

fungi

Mode of action

DNA synthesis | interferes
enzyme | inhibits

storage temp.

−20°C

SMILES string

CO[C@H]1\C=C\C=C(C)\C[C@H](C)[C@H](O)[C@H](C)\C=C(C)\C=C(OC)C(=O)OC1[C@@H](C)[C@@H](O)[C@H](C)[C@@]2(O)C[C@@H](O)[C@H](C)[C@H](O2)C(C)C

InChI

1S/C35H58O9/c1-19(2)32-24(7)27(36)18-35(40,44-32)26(9)31(38)25(8)33-28(41-10)14-12-13-20(3)15-22(5)30(37)23(6)16-21(4)17-29(42-11)34(39)43-33/h12-14,16-17,19,22-28,30-33,36-38,40H,15,18H2,1-11H3/b14-12+,20-13+,21-16+,29-17-/t22-,23+,24-,25-,26-,27+,28-,30-,31+,32+,33+,35+/m0/s1

InChI key

XDHNQDDQEHDUTM-JQWOJBOSSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: macrolide

Application

Bafilomycin A1 has been used:
  • as an autophagy inhibitor to study its effects on hepatic steatosis in human hepatocytes
  • as a vacuolar-type H+ -ATPase inhibitor to study its effects on the cell viability, percentage of PI-positive cells, and glucose-stimulated insulin secretion (GSIS) in mice INS-1 cells
  • as a vacuolar-type H+ -ATPase inhibitor to study its effects on tau degradation in human SH-SY5Y cells

Biochem/physiol Actions

A specific inhibitor of vacuolar type H+-ATPase (V-ATPase) in animal cells, plant cells and microorganisms.
Bafilomycin A1 inhibits autophagosome-lysosome fusion and autolysosome acidification, the steps involved in the autophagic process that is required for maintaining functional autophagic flux and cellular homeostasis.
Specific inhibitor of vacuolar type H+-ATPase (V-ATPase).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jens Niewoehner et al.
Neuron, 81(1), 49-60 (2014-01-15)
Although biotherapeutics have vast potential for treating brain disorders, their use has been limited due to low exposure across the blood-brain barrier (BBB). We report that by manipulating the binding mode of an antibody fragment to the transferrin receptor (TfR)
ERK-dependent mTOR pathway is involved in berberine-induced autophagy in hepatic steatosis
He Q, et al.
Journal of Molecular Endocrinology, 57(4) (2016)
Ru Li et al.
Phytopathology, 109(8), 1417-1424 (2019-03-13)
The vacuolar H+-ATPases (V-ATPases) are conserved ATP-dependent proton pumps that acidify intracellular compartments in eukaryotic cells. The role of Cpvma1, a V-ATPase catalytic subunit A of Cryphonectria parasitica, was investigated by generating cpvma1-overexpressing and cpvma1-silenced strains. The mutant strains were
Caroline Mauvezin et al.
Autophagy, 11(8), 1437-1438 (2015-07-15)
Autophagosome-lysosome fusion and autolysosome acidification constitute late steps in the autophagic process necessary to maintain functional autophagic flux and cellular homeostasis. Both of these steps are disrupted by the V-ATPase inhibitor bafilomycin A1, but the mechanisms potentially linking them are
Bastien Mangeat et al.
PLoS pathogens, 5(9), e1000574-e1000574 (2009-09-05)
Host cells impose a broad range of obstacles to the replication of retroviruses. Tetherin (also known as CD317, BST-2 or HM1.24) impedes viral release by retaining newly budded HIV-1 virions on the surface of cells. HIV-1 Vpu efficiently counteracts this

Articles

We presents an article on Autophagy in Cancer Promotes Therapeutic Resistance

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service