All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C27H40N8O6
CAS Number:
Molecular Weight:
572.66
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32
Recommended Products
Assay
≥97% (HPLC)
form
powder
UniProt accession no.
storage temp.
−20°C
SMILES string
NC(CCCNC(N)=N)C(=O)N1CCCC1C(=O)N2CCCC2C(=O)NCC(=O)NC(Cc3ccccc3)C(O)=O
InChI
1S/C27H40N8O6/c28-18(9-4-12-31-27(29)30)24(38)35-14-6-11-21(35)25(39)34-13-5-10-20(34)23(37)32-16-22(36)33-19(26(40)41)15-17-7-2-1-3-8-17/h1-3,7-8,18-21H,4-6,9-16,28H2,(H,32,37)(H,33,36)(H,40,41)(H4,29,30,31)
InChI key
USSUMSBPLJWFSZ-UHFFFAOYSA-N
Gene Information
human ... KNG1 (includes EG:3827)
Related Categories
Amino Acid Sequence
Arg-Pro-Pro-Gly-Phe
Biochem/physiol Actions
The bradykinin (BK) fragment (1-5) (RPPGF) is among the most stable of naturally occurring metabolites. It may be used as a marker for BK production in vivo. It is known that an intact Arg residue in the C-terminus is required for biological activities. BK 1-5 is the minimal peptide that inhibited α-thrombin-induced platelet aggregation and secretion and calcium mobilization. It also prevented α-thrombin from cleaving the thrombin receptor peptide, NATLDPRSFLLR, between arginine and serine. Such antithrombin activities of BK 1-5 may contribute to the cardioprotective nature of kinins.
Packaging
Bottomless glass bottle. Contents are inside inserted fused cone.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
A A Hasan et al.
Thrombosis and haemostasis, 82(3), 1182-1187 (1999-09-24)
Thrombostatin (RPPGF), an angiotensin converting enzyme metabolite of bradykinin, is an inhibitor of alpha-thrombin's ability to activate platelets. We examined the in vivo pharmacokinetics and pharmacodynamics of thrombostatin in rabbits and its ability to inhibit coronary thrombosis induced by electrolytic
Marvin T Nieman et al.
The Journal of pharmacology and experimental therapeutics, 311(2), 492-501 (2004-06-24)
Thrombin and protease-activated receptor 1 (PAR1) activation antagonists were prepared based upon the peptide RPPGF, the angiotensin-converting enzyme breakdown product of bradykinin. A library of 72 peptides consisting of d and/or synthetic amino acids was designed with various substitutions in
T A Morinelli et al.
The Journal of pharmacology and experimental therapeutics, 296(1), 71-76 (2000-12-21)
Extensive research has provided few therapeutic agents for the treatment of septicemia. Bradykinin, an endogenous vasodepressor hormone, is a key mediator in the hypotension seen with septicemia. The present investigation shows that a stable metabolic fragment of bradykinin, arginine-proline-proline-glycine-phenylalanine (RPPGF)
Cho Yeow Koh et al.
Thrombosis and haemostasis, 102(3), 437-453 (2009-09-01)
To obtain blood meals, haematophagous animals are armed with potent pharmacological molecules to overcome several of their hosts' response systems. Among them, a large number of exogenous anticoagulants which target the haemostatic system have been identified and characterised. Studies on
L J Murphey et al.
Circulation, 102(8), 829-832 (2000-08-23)
Bradykinin is a cardioprotective peptide metabolized by the angiotensin-converting enzyme (ACE). An insertion/deletion (I/D) polymorphism in the ACE gene determines plasma ACE levels. The D allele is associated with cardiovascular disease, which may relate to enhanced angiotensin II production or
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service