Skip to Content
Merck
All Photos(1)

Key Documents

05-23-0101

Sigma-Aldrich

Angiotensin II, Human

Plays an important role in the regulation of blood pressure.

Synonym(s):

Angiotensin II, Human, DRVYIHPF, ang II

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C50H71N13O12
CAS Number:
Molecular Weight:
1046.18
UNSPSC Code:
12352202
NACRES:
NA.77

biological source

human

Quality Level

sterility

non-sterile

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

technique(s)

cell based assay: suitable

color

white to off-white

solubility

5% acetic acid: 1 mg/mL
water: 1 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

CCC(C)C(C(=O)NC(CC1=CN=CN1)C(=O)N2CCCC2C(=O)NC(CC3=CC=CC=C3)C(=O)O)NC(=O)C(CC4=CC=C(C=C4)O)NC(=O)C(C(C)C)NC(=O)C(CCCN=C(N)N)NC(=O)C(CC(=O)O)N

InChI

1S/C50H71N13O12/c1-5-28(4)41(47(72)59-36(23-31-25-54-26-56-31)48(73)63-20-10-14-38(63)45(70)60-37(49(74)75)22-29-11-7-6-8-12-29)62-44(69)35(21-30-15-17-32(64)18-16-30)58-46(71)40(27(2)3)61-43(68)34(13-9-19-55-50(52)53)57-42(67)33(51)24-39(65)66/h6-8,11-12,15-18,25-28,33-38,40-41,64H,5,9-10,13-14,19-24,51H2,1-4H3,(H,54,56)(H,57,67)(H,58,71)(H,59,72)(H,60,70)(H,61,68)(H,62,69)(H,65,66)(H,74,75)(H4,52,53,55)/t28-,33-,34-,35-,36-,37-,38-,40-,41?/m0/s1

InChI key

CZGUSIXMZVURDU-GYGDIBSWSA-N

Gene Information

human ... ACE2(59272)

General description

Research area: Cell signaling

Cell permeable: no

Primary Target adenylate cyclase activity

Product does not compete with ATP.

Reversible: no

Application

Angiotensin II, Human has been used to induce fibrosis in mice to evaluate the impact of fibroblast loss on cardiac function after injury. It has also been used to induce abdominal aortic aneurysm (AAA) progression in mice.

Biochem/physiol Actions

Angiotensin II is a crucial component of the Renin-Angiotensin System (RAS). It plays a vital role in vasoconstriction, regulating blood pressure, and controlling fluid balance by influencing the retention of sodium and water in the renal tubules. Furthermore, Angiotensin II stimulates the release of aldosterone from the adrenal gland, contributing to the maintenance of fluid and electrolyte balance regulation. Angiotensin II also promotes inflammation and tissue injury. It Inhibits adenylate cyclase activity in spontaneously hypertensive rats. Angiotensin II also activates JAK2 in smooth muscle cells, as well as several kinases including p125FAK, cytosolic calcium-dependent tyrosine kinase, and p60c-src in various cell types. In vascular smooth muscle cells, it activates ERK1/2, JNK, and p38 MAP Kinase.

Packaging

Yes

Warning

Toxicity: Standard Handling (A)

Sequence

H-Asp-Arg-Val-Tyr-Ile-His-Pro-Phe-OH

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 week at 4°C or for up to 1 month at -20°C.

Other Notes

Zhu, M., et al. 2000. J. Neurophysiol.84, 2494.
Natarajan, R., et al. 1999. Hypertension 33, 378.
Mazzolai, L., et al. 1998. Hypertension 31, 1324.
Earp, H.S., et al. 1995. J. Biol. Chem. 270, 28440.
Ishida, M., et al. 1995. Circ. Res. 77, 1053.
Vyas, S.J., and Jackson, E.K. 1995. J. Pharmacol. Exp. Ther. 273, 768.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Early Progression of Abdominal Aortic Aneurysm is Decelerated by Improved Endothelial Barrier Function via ALDH2-LIN28B-ELK3 Signaling
Yang K, et al.
Journal of Separation Science, 10(32) (2023)
Activation of MAPKs by angiotensin II in vascular smooth muscle cells. Metalloprotease-dependent EGF receptor activation is required for activation of ERK and p38 MAPK but not for JNK
Eguchi S, et al.
The Journal of Biological Chemistry, 276(11), 7957-7962 (2001)
Angiotensin II inhibits glomerular adenylate cyclase via the angiotensin II receptor subtype 1 (AT1)
Edwards R M and Stack E J
The Journal of Pharmacy and Pharmacology, 266(2), 506-510 (1993)
Early Progression of Abdominal Aortic Aneurysm is Decelerated by Improved Endothelial Barrier Function via ALDH2-LIN28B-ELK3 Signaling
Yang K, et al.
Advanced Science Letters, 10(32) (2023)
Angiotensin II revisited: new roles in inflammation, immunology and aging
Benigni A, et al.
EMBO Molecular Medicine, 2(7), 247-257 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service