Skip to Content
Merck
All Photos(1)

Key Documents

A9650

Sigma-Aldrich

Angiotensin I human acetate salt hydrate

≥90% (HPLC), powder, suitable for mammalian cell culture

Synonym(s):

5-Ile-angiotensin I, 5-Isoleucine-angiotensin I, Pepsitensin, Proangiotensin, Hypertensin I

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C62H89N17O14 · xC2H4O2 · yH2O
CAS Number:
Molecular Weight:
1296.48 (anhydrous free base basis)
MDL number:
UNSPSC Code:
12352202
eCl@ss:
42020611
PubChem Substance ID:
NACRES:
NA.32

biological source

human

Quality Level

Assay

≥90% (HPLC)

form

powder

mol wt

1296 Da by calculation (Average Mass)

storage condition

(Keep container tightly closed in a dry and well-ventilated place)

technique(s)

cell culture | mammalian: suitable

solubility

acetic acid: 25 mg/mL, clear, colorless

UniProt accession no.

storage temp.

−20°C

SMILES string

CC(O)=O.CC(O)=O.CC[C@H](C)[C@H](NC(=O)[C@H](Cc1ccc(O)cc1)NC(=O)[C@@H](NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@@H](N)CC(O)=O)C(C)C)C(=O)N[C@@H](Cc2c[nH]cn2)C(=O)N3CCC[C@H]3C(=O)N[C@@H](Cc4ccccc4)C(=O)N[C@H](Cc5c[nH]cn5)C(=O)N[C@@H](CC(C)C)C(O)=O

InChI

1S/C62H89N17O14.2C2H4O2/c1-7-35(6)51(78-56(87)44(25-37-17-19-40(80)20-18-37)74-58(89)50(34(4)5)77-53(84)42(15-11-21-68-62(64)65)71-52(83)41(63)28-49(81)82)59(90)75-46(27-39-30-67-32-70-39)60(91)79-22-12-16-48(79)57(88)73-43(24-36-13-9-8-10-14-36)54(85)72-45(26-38-29-66-31-69-38)55(86)76-47(61(92)93)23-33(2)3;2*1-2(3)4/h8-10,13-14,17-20,29-35,41-48,50-51,80H,7,11-12,15-16,21-28,63H2,1-6H3,(H,66,69)(H,67,70)(H,71,83)(H,72,85)(H,73,88)(H,74,89)(H,75,90)(H,76,86)(H,77,84)(H,78,87)(H,81,82)(H,92,93)(H4,64,65,68);2*1H3,(H,3,4)/t35-,41-,42-,43-,44-,45+,46-,47-,48-,50-,51-;;/m0../s1

InChI key

CKIFOTYDIFUKEP-AUVUXLIHSA-N

Gene Information

human ... AGT(183)

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Asp-Arg-Val-Tyr-Ile-His-Pro-Phe-His-Leu

General description

Angiotensinogen (Angiotensin I) is a renin substrate, which is produced by the liver.

Research area: Cell Signaling

Application

Angiotensin I human acetate salt hydrate has been used:
  • as an external calibration standard, it was applied to α-cyano-4-hydroxycinnamic acid (CHCA) and 2,5-dihydroxybenzoic acid (DHB) matrix,
  • to prepare angiotensin I solution gravimetrically for hydrolysis and also used as a model peptide to demonstrate SI (système international) -traceable amino acid analysis
  • as an analyte to deposit on the target in the study of on-line single droplet deposition for matrix-assisted laser desorption/ionization (MALDI) mass spectrometry

Biochem/physiol Actions

Angiotensin I is a putative neurotransmitter. It is the precursor of angiotensin II and of angiotensin fragment 1-7, which are the hormones involved in regulation of fluid volume and the release of aldosterone.
Angiotensin is a component of the renin-angiotensin aldosterone system (RAAS). This hormonal circulating system aids in regulating blood pressure by modulating potassium secretion, vascular tone, and sodium and water reabsorption. RAAS also participates in fibrosis, apoptosis, and inflammation.
Putative neurotransmitter; precursor of angiotensin II and of angiotensin fragment 1-7.

Antibody Adsorbant

Asp-Arg-Val-Tyr-Ile-His-Pro-Phe-His-Leu-OH.2CH3COOH.6H2O

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R A Santos et al.
Regulatory peptides, 91(1-3), 45-62 (2000-09-01)
The renin-angiotensin system is a major physiological regulator of arterial pressure and hydro-electrolyte balance. Evidence has now been accumulated that in addition to angiotensin (Ang) II other Ang peptides [Ang III, Ang IV and Ang-(1-7)], formed in the limited proteolysis
Toward Systeme International d?Unite-traceable protein quantification: from amino acids to proteins
Burkitt W I, et al.
Analytical biochemistry, 376(2), 242-251 (2008)
Genetics of Blood Pressure Regulation
Emery and Rimoin's Principles and Practice of Medical Genetics, 1-22 (2013)
Physiology, Renin Angiotensin System
Fountain J H. and Lappin S L.
StatPearls [Internet] (2022)
Protein expression following heat shock in the nervous system of Locusta migratoria
Dehghani M, et al.
Journal of Insect Physiology, 57(11), 1480-1488 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service