Skip to Content
Merck
All Photos(1)

Key Documents

B1901

Sigma-Aldrich

Bradykinin Fragment 2-9

≥97% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H61N11O10
CAS Number:
Molecular Weight:
904.02
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32

Assay

≥97% (HPLC)

form

powder

UniProt accession no.

storage temp.

−20°C

SMILES string

NC(=N)NCCCC(NC(=O)C(Cc1ccccc1)NC(=O)C2CCCN2C(=O)C(CO)NC(=O)C(Cc3ccccc3)NC(=O)CNC(=O)C4CCCN4C(=O)C5CCCN5)C(O)=O

InChI

1S/C44H61N11O10/c45-44(46)48-20-8-16-30(43(64)65)51-38(59)32(24-28-13-5-2-6-14-28)52-40(61)35-18-10-22-55(35)42(63)33(26-56)53-37(58)31(23-27-11-3-1-4-12-27)50-36(57)25-49-39(60)34-17-9-21-54(34)41(62)29-15-7-19-47-29/h1-6,11-14,29-35,47,56H,7-10,15-26H2,(H,49,60)(H,50,57)(H,51,59)(H,52,61)(H,53,58)(H,64,65)(H4,45,46,48)

InChI key

ZZHVXIPXTCBVBE-UHFFFAOYSA-N

Gene Information

Amino Acid Sequence

Pro-Pro-Gly-Phe-Ser-Pro-Phe-Arg

Biochem/physiol Actions

Inhibits ACE (angiotensin converting enzyme) and potentiates bradykinin (BK).
The des-arginine-1 bradykinin (BK) peptide (2-9) has no detectable contractile activity, inhibits ACE (angiotensin converting enzyme) and potentiates BK.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B W Busser et al.
The Journal of urology, 160(6 Pt 1), 2267-2273 (1998-11-17)
Bradykinin 1 (B1) receptors have been shown to be upregulated at sites of inflammation. The purpose of this study was to determine the effect of lipopolysaccharide (LPS) on B1 receptor modulation in the isolated mouse bladder. The contractile responses of
Cristiano R Jesse et al.
Pharmacology, biochemistry, and behavior, 91(4), 573-580 (2008-10-22)
The objective of this study was to extend our previous findings by investigating in greater detail the mechanisms that might be involved in the antinociceptive action of p-methoxyl-diphenyl diselenide, (MeOPhSe)(2), in mice. The pretreatment with nitric oxide precursor, l-arginine (600
S Mohammed et al.
Journal of mass spectrometry : JMS, 36(12), 1260-1268 (2001-12-26)
A novel tandem quadrupole mass spectrometer is described that enables gaseous collision-induced dissociation (CID) and surface-induced dissociation (SID) experiments. The instrument consists of a commercially available triple quadrupole mass spectrometer connected to an SID region and an additional, orthogonal quadrupole
G S Bedi et al.
Biochimica et biophysica acta, 842(1), 90-99 (1985-09-27)
Four hybridoma cell lines have been established that secrete monoclonal antibodies to nonapeptide bradykinin. Bradykinin coupled to ovalbumin, using 1-ethyl-3-(3-dimethylaminopropyl)carbodiimide as coupling agent, was used to immunize BALB/c mice. Spleen cells from the immunized animals were fused to P3-X63-AG8-653 mouse
Julia Laskin et al.
Journal of the American Chemical Society, 125(6), 1625-1632 (2003-02-06)
Time- and collision energy-resolved surface-induced dissociation (SID) of des-Arg(1)- and des-Arg(9)-bradykinin on a fluorinated self-assembled monolayer (SAM) surface was studied by use of a novel Fourier transform ion cyclotron resonance mass spectrometer (FT-ICR MS) specially equipped to perform SID experiments.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service