Skip to Content
Merck
All Photos(1)

Key Documents

N5511

Supelco

(−)-Nicotine solution

1.0 mg/mL, analytical standard, for drug analysis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14N2
CAS Number:
Molecular Weight:
162.23
Beilstein:
5733110
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard, for drug analysis

Quality Level

concentration

1.0 mg/mL in methanol
1.0 mg/mL

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
cleaning products
cosmetics
food and beverages
personal care
pharmaceutical (small molecule)

format

single component solution

storage temp.

2-8°C

SMILES string

CN1CCC[C@H]1c2cccnc2

InChI

1S/C10H14N2/c1-12-7-3-5-10(12)9-4-2-6-11-8-9/h2,4,6,8,10H,3,5,7H2,1H3/t10-/m0/s1

InChI key

SNICXCGAKADSCV-JTQLQIEISA-N

Looking for similar products? Visit Product Comparison Guide

Application

(−)-Nicotine solution has been used as a reference standard in the analysis of nicotine in air affected by second hand smoke using ultra performance liquid chromatography (UPLC).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Assessment of nicotine for second hand smoke exposure in some public places in Romania by UPLC-MS
Bratan A, et al.
Chemistry Central Journal, 8(1), 29-29 (2014)
E-cigarettes, smokers and health policies.
S Nardini et al.
Monaldi archives for chest disease = Archivio Monaldi per le malattie del torace, 79(1), 6-7 (2013-06-08)
Jonathan P Winickoff et al.
Pediatrics, 132(1), 109-117 (2013-06-26)
To test whether routine pediatric outpatient practice can be transformed to assist parents in quitting smoking. Cluster RCT of 20 pediatric practices in 16 states that received either CEASE intervention or usual care. The intervention gave practices training and materials
The regulatory challenge of electronic cigarettes.
Neal L Benowitz et al.
JAMA, 310(7), 685-686 (2013-07-17)
Governments must agree unified approach to use of e-cigarettes, report says.
Susan Mayor
BMJ (Clinical research ed.), 346, f3537-f3537 (2013-06-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service