Skip to Content
Merck
All Photos(1)

Documents

N3876

Sigma-Aldrich

(−)-Nicotine

≥99% (GC), liquid

Synonym(s):

(−)-1-Methyl-2-(3-pyridyl)pyrrolidine, (S)-3-(1-Methyl-2-pyrrolidinyl)pyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14N2
CAS Number:
Molecular Weight:
162.23
Beilstein:
82109
EC Number:
MDL number:
UNSPSC Code:
12352210
PubChem Substance ID:
NACRES:
NA.77

Assay

≥99% (GC)

form

liquid

optical activity

[α]20/D −169°(lit.)

color

yellow

refractive index

n20/D 1.5265 (lit.)

solubility

ethanol: 50 mg/mL

density

1.010 g/mL at 20 °C (lit.)

SMILES string

CN1CCC[C@H]1c2cccnc2

InChI

1S/C10H14N2/c1-12-7-3-5-10(12)9-4-2-6-11-8-9/h2,4,6,8,10H,3,5,7H2,1H3/t10-/m0/s1

InChI key

SNICXCGAKADSCV-JTQLQIEISA-N

Looking for similar products? Visit Product Comparison Guide

General description

Nicotine is the most abundant alkaloid found in the leaves of Nicotiana species. It is a major toxic agent.

Application

(-)-Nicotine has been used:
  • to stimulate neutrophils in extracellular DNA fluorescence assay and in immunostaining of neutrophils
  • to study the effects of chronic neonatal nicotine exposure on nicotinic acetylcholine receptor (nAChR) binding, cell death and morphology in hippocampus and cerebellum
  • in an in vitro study to evaluate the effect of root exposure to nicotine or cotinine on the morphology and density of fibroblasts

Biochem/physiol Actions

Nicotine binds to nicotinic acetylcholine receptors and exhibit both neuroprotective and neurotoxic effects on the developing brain. It is associated with lung cancer development and reduces the efficiency of chemotherapeutic agents. In addition, nicotine has an ability to impair phagocytic host defense and also increase reactive oxygen species (ROS) associated injury. It also stops Akt/ protein kinase B (PKB) deactivation and decreases spontaneous death of neutrophils. Nicotine is used to treat Alzheimer′s and Parkinson′s disease.
Prototype nicotinic acetylcholine receptor agonist; naturally occurring isomer.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Oral - Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

213.8 °F - closed cup

Flash Point(C)

101 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Effects of chronic neonatal nicotine exposure on nicotinic acetylcholine receptor binding, cell death and morphology in hippocampus and cerebellum
Huang LZ, et al.
Neuroscience, 146(4), 1854-1868 (2007)
Nicotine alkaloid levels, and nicotine to nornicotine conversion, in Australian Nicotiana species used as chewing tobacco
Moghbel N, et al.
Heliyon, 3(11), e00469-e00469 (2017)
Nicotine induces neutrophil extracellular traps
Hosseinzadeh A, et al.
Journal of Leukocyte Biology, 100(5), 1105-1112 (2016)
Root surface conditioning with nicotine or cotinine reduces viability and density of fibroblasts in vitro
Martinez AE, et al.
Clinical Oral Investigations, 9(3), 180-186 (2005)
Nicotine inactivation of the proapoptotic function of Bax through phosphorylation
Xin M and Deng X
The Journal of Biological Chemistry, 280(11), 10781-10789 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service