Skip to Content
Merck
All Photos(1)

Key Documents

N-008

Supelco

S(−)-Nicotine solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

(−)-Nicotine solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14N2
CAS Number:
Molecular Weight:
162.23
Beilstein:
5733110
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

CN1CCC[C@H]1c2cccnc2

InChI

1S/C10H14N2/c1-12-7-3-5-10(12)9-4-2-6-11-8-9/h2,4,6,8,10H,3,5,7H2,1H3/t10-/m0/s1

InChI key

SNICXCGAKADSCV-JTQLQIEISA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

A Certified Spiking Solution® applicable for use in LC/MS or GC/MS applications such as nicotine testing, forensic analysis or clinical and diagnostic testing. Nicotine is a highly additive tobacco alkaloid associated with numerous health risks such as heart health, mental health and cancer.

Application


  • Chiral nicotine for neuroreceptor research: S(−)-Nicotine solution is extensively used in neuroreceptor studies, particularly targeting cholinergic receptors. This enantiopure nicotine variant helps in understanding the binding affinity and activity differences between nicotine isomers, contributing to more targeted neuropharmacological interventions (Lu and Gao, 2024).

  • Pharmacological experiments in addiction research: The optically active form of S(−)-Nicotine is critical for addiction research, where it is used to investigate the pharmacodynamics and addictive properties of nicotine. Studies using this compound help in developing therapeutic strategies for smoking cessation and understanding the biochemical pathways involved in nicotine addiction (Yang et al., 2024).

  • Enantiomer specificity in biochemical studies: High purity S(−)-Nicotine is crucial for assays that require precise control over biochemical properties. Its application in cholinergic receptor assays helps to elucidate the specific interactions and effects of the S(−) enantiomer compared to its R(+) counterpart, thereby enhancing the accuracy of pharmacological models (Mancuso et al., 2024).

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jia Gao et al.
Cell death & disease, 12(6), 508-508 (2021-05-20)
There is limited and discordant evidence on the role of nicotine in diabetic vascular disease. Exacerbated endothelial cell dysregulation in smokers with diabetes is associated with the disrupted adipose function. Adipokines possess vascular protective, anti-inflammatory, and anti-diabetic properties. However, whether
Muhammed Ershad et al.
Journal of medical toxicology : official journal of the American College of Medical Toxicology, 16(4), 452-457 (2020-03-18)
Cigarettes and other tobacco products may be extinguished by submersion in liquids in beverage cans or bottles. Cases of nicotine poisoning in children have been reported following ingestion of such liquids. The aim of this study is to analyze the
Delia Dempsey et al.
Clinical pharmacology and therapeutics, 76(1), 64-72 (2004-07-02)
Nicotine and a variety of other drugs and toxins are metabolized by cytochrome P450 (CYP) 2A6. Our objective was to evaluate the use of oral nicotine with measurement of the trans-3'-hydroxycotinine (3HC)/cotinine (COT) metabolite ratio as a noninvasive probe of
S Miksys et al.
Journal of neural transmission. Supplementum, (70)(70), 177-180 (2006-10-05)
Brain expression of cytochromes P450 2B6, 2D6 and 2E1 is higher in smokers, and is induced by nicotine in animals. These enzymes can metabolize many of the neurotoxins associated with Parkinson's disease. Since smoking is known to be protective against
Svetlana Farkas et al.
Toxicology and applied pharmacology, 214(2), 118-125 (2006-02-14)
Epidemiological studies support an association between perinatal cigarette smoke (CS) exposure and a number of severe pre- and postnatal complications. However, the mechanisms through which CS enhances such risks largely remain unknown. One of the reasons for our inability to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service