Skip to Content
Merck
All Photos(1)

Key Documents

N0267

Sigma-Aldrich

(±)-Nicotine

≥99% (TLC), liquid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14N2
CAS Number:
Molecular Weight:
162.23
Beilstein:
82108
MDL number:
UNSPSC Code:
12352210
eCl@ss:
39151701
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥99% (TLC)

form

liquid

optical activity

[α]25/D 0°(lit.)

color

yellow

solubility

ethanol: 50 mg/mL

density

1.02 g/mL at 25 °C (lit.)

SMILES string

CN1CCCC1c2cccnc2

InChI

1S/C10H14N2/c1-12-7-3-5-10(12)9-4-2-6-11-8-9/h2,4,6,8,10H,3,5,7H2,1H3

InChI key

SNICXCGAKADSCV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

(±)-Nicotine has been used:
  • as a supplement of high-glucose Dulbecco′s modified Eagle′s medium, to test its effect on mesenchymal stem cells
  • as a standard for the determination of nicotine concentration using phase high-pressure liquid chromatography
  • as a test compound to study its effects on fibril formation

Biochem/physiol Actions

(±)-Nicotine is a tertiary amine compound that is predominantly present in tobacco. It binds to the nicotinic cholinergic receptors of the brain, autonomic ganglia and neuromuscular junction. Nicotine allosterically regulates a number of biological events such as production of growth hormone, dopamine, adrenocorticotropic hormone and prolactin. Nicotine absorption across biological membranes is pH sensitive, which alters its ionization state. Absorption of nicotine is shown to be higher at alkaline pH, when it remains unionized. Nicotine metabolism occurs extensively in the liver and it can cross placenta.
Prototype nicotinic acetylcholine receptor agonist.

Packaging

The material is liquid, transparent, and viscous, packaged in amber vail, and the volume inserted into each bottle is very small, It may form a thin FILM on the sides. While it may be difficult to see the material inside by the eyes, the container is not empty.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

213.8 °F - closed cup

Flash Point(C)

101 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

The effect of nicotine on the mechanical properties of mesenchymal stem cells
Ruiz JP, et al.
Cell health and cytoskeleton, 4, 29-29 (2012)
Guangxin Luan et al.
Experimental and therapeutic medicine, 23(1), 11-11 (2021-11-25)
Pulmonary emphysema is one of the most important pathological manifestations of chronic obstructive pulmonary disease and is commonly associated with cigarette smoking. Previous studies have indicated that necroptosis, a novel non-apoptotic cell death mechanism associated with inflammation and oxidative stress
Neal L Benowitz
Nicotine Safety and Toxicity, 3-7 (1998)
Alterations in the production and concentration of selected alkaloids as a function of rising atmospheric carbon dioxide and air temperature: implications for ethno-pharmacology
Ziska LH, et al.
Global Change Biology, 11(10), 1798-1807 (2005)
Daniel A Kirschner et al.
Current Alzheimer research, 5(3), 288-307 (2008-06-10)
Targeting the initial formation of amyloid assemblies is a preferred approach to therapeutic intervention in amyloidoses, which include such diseases as Alzheimer's, Parkinson's, Huntington's, etc., as the early-stage, oligomers that form before the development of beta-conformation-rich fibers are thought to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service