Skip to Content
Merck
All Photos(3)

Key Documents

919780

Sigma-Aldrich

[1,1′-Bis(diphenylphosphino)ferrocene]dichloropalladium(II) ChemBeads

Synonym(s):

Pd(dppf)Cl2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C17H14P)2Fe . PdCl2
CAS Number:
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.22

form

solid

Quality Level

composition

~ 4 wt.% loading of catalyst

reaction suitability

reagent type: catalyst
reaction type: Cross Couplings

SMILES string

[Pd+2].[Fe+2].P(c6c[c-H]cc6)(c5ccccc5)c4ccccc4.P(c3c[c-H]cc3)(c2ccccc2)c1ccccc1.[Cl-].[Cl-]

InChI

1S/2C17H14P.2ClH.Fe.Pd/c2*1-3-9-15(10-4-1)18(17-13-7-8-14-17)16-11-5-2-6-12-16;;;;/h2*1-14H;2*1H;;/q2*-1;;;2*+2/p-2

InChI key

NXQGGXCHGDYOHB-UHFFFAOYSA-L

Application

Catalyst for C-C and C-N coupling reaction.
Pd(dppf)Cl2 may be used as an effective palladium catalyst in the following reactions:
  • Cross-coupling of sec-alkyl and n-alkyl Grignard reagents with high yield and selectivity.
  • Suzuki coupling of aryl boronic esters with [11C]methyl iodide to form functionalized [11C]toluene derivatives.
  • Kumada cross-coupling of 1,3,5-tribromobenzene with Grignard reagents to form star-shaped oligothiophenes.

ChemBeads are chemical coated glass beads. ChemBeads offer improved flowability and chemical uniformity perfect for automated solid dispensing and high-throughput experimentation. The method of creating ChemBeads uses no other chemicals or surfactants allowing the user to accurately dispense sub-milligram amounts of chemical.

Learn more about ChemBeads products

For larger scale uses, product also available in powdered form (697230) & (931217)

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B Inhalation

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Star−Shaped Oligothiophenes for Solution−Processible Organic Field-Effect Transistors
Ponomarenko S A, et al.
Advances in Functional Materials, 13(8), 591-596 (2003)
An improved synthesis of substituted [11C]toluenes via Suzuki coupling with [11C]methyl iodide
Hostetler E, et al.
Journal of Labelled Compounds & Radiopharmaceuticals, 48(9(, 629-634 (2005)
Dichloro[1,1′-bis(diphenylphosphino)ferrocene]palladium(II): an effective catalyst for cross-coupling reaction of a secondary alkyl grignard reagent with organic halides
Hayashi T, et al.
Tetrahedron Letters, 20(21), 1871-1874 (1979)
Versatile Methods to Dispense Sub-Milligram Quantities of Solids using Chemical Coated Beads for High-Throughput Experimentation.
Martin C M, et al.
Organic Process Research & Development, 23(9), 1900-1907 (2019)
Sho Fujii et al.
Physical chemistry chemical physics : PCCP, 19(42), 28943-28949 (2017-10-24)
A novel sensor, 4-[2-(9-anthryl)ethynyl]-1,10-phenanthroline (1), exhibits highly intense fluorescent in the wavelength region of 440-600 nm (maximum wavelength (λf) = 470 nm) with the quantum yield (Φf) and lifetime (τf) being 0.90 and 4.2 ns, respectively, in CH3CN at 298

Articles

ChemBeads, catalyst-coated glass beads, dispense solid chemical reagents for nanomole-scale high-throughput reaction screening.

ChemBeads, catalyst-coated glass beads, dispense solid chemical reagents for nanomole-scale high-throughput reaction screening.

ChemBeads, catalyst-coated glass beads, dispense solid chemical reagents for nanomole-scale high-throughput reaction screening.

ChemBeads, catalyst-coated glass beads, dispense solid chemical reagents for nanomole-scale high-throughput reaction screening.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service