Skip to Content
Merck
All Photos(6)

Key Documents

227994

Sigma-Aldrich

Bis(dibenzylideneacetone)palladium(0)

Synonym(s):

Palladium(0) bis(dibenzylideneacetone), Pd(dba)2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5CH=CHCOCH=CHC6H5)2Pd
CAS Number:
Molecular Weight:
575.00
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

form

powder

Quality Level

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Cross Couplings
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

SMILES string

[Pd].O=C(/C=C/c1ccccc1)\C=C\c2ccccc2.O=C(/C=C/c3ccccc3)\C=C\c4ccccc4

InChI

1S/2C17H14O.Pd/c2*18-17(13-11-15-7-3-1-4-8-15)14-12-16-9-5-2-6-10-16;/h2*1-14H;/b2*13-11+,14-12+;

InChI key

UKSZBOKPHAQOMP-SVLSSHOZSA-N

General description

Bis(dibenzylideneacetone)palladium(0) (Pd(dba)2) is an air stable Pd0 complex. It is a reagent for the synthesis of allylic substituted cyclopentadienes. It is a homogeneous catalyst. It catalyzes the alkylation of allyl acetates by various nucleophiles under mild conditions.

Application

Bis(dibenzylideneacetone)palladium(0) (Pd(dba)2) was employed as catalyst in the following studies:
  • Synthesis of isomeric 2-aryl-2,5-dihydrofurans, via Heck coupling of aryl bromides with alkenes using neopentyl phosphine ligands.
  • Heck reaction of benzyl trifluoroacetate and 2,3-dihydrofuran phosphoramidite ligand.
  • Allylation of stabilized anions.
  • Cross coupling of allyl, alkenyl and aryl halides with organostannanes.
  • Cross coupling of vinyl halides with alkenyl zinc species.
  • Carbonylation of alkenyl and aryl halides.
  • Employed with cyclic thiourea ligands in an efficient aerobic oxidation of alcohols to aldehydes and ketones.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Macsari, I.; Hupe, E.; Szabo, K. J.
The Journal of Organic Chemistry, 64, 9547-9547 (1999)
T Vats et al.
Scientific reports, 6, 33053-33053 (2016-09-14)
Amazing conductivity, perfect honeycomb sp(2) arrangement and the high theoretical surface area make pristine graphene as one of the best materials suited for application as catalyst supports. Unfortunately, the low reactivity of the material makes the formation of nanocomposite with
Arentsen, K.; Caddick, S.et al.
Tetrahedron Letters, 45, 3511-3511 (2004)
Tetrahedron Letters, 25, 4379-4379 (1984)
Tetrahedron Letters, 21, 4437-4437 (1980)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service