Skip to Content
Merck
All Photos(4)

Documents

379670

Sigma-Aldrich

[1,1′-Bis(diphenylphosphino)ferrocene]dichloropalladium(II), complex with dichloromethane

Synonym(s):

Pd(dppf)Cl2 · CH2Cl2, Pd(dppf)Cl2 · DCM, [1,1′-Bis(diphenylphosphino)ferrocene]dichloropalladium(II) (1:1)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H30Cl4FeP2Pd
CAS Number:
Molecular Weight:
816.64
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

form

solid

Quality Level

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Cross Couplings
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

mp

275-280 °C

SMILES string

[Fe].ClCCl.Cl[Pd]Cl.[CH]1[CH][CH][C]([CH]1)P(c2ccccc2)c3ccccc3.[CH]4[CH][CH][C]([CH]4)P(c5ccccc5)c6ccccc6

InChI

1S/2C17H14P.CH2Cl2.2ClH.Fe.Pd/c2*1-3-9-15(10-4-1)18(17-13-7-8-14-17)16-11-5-2-6-12-16;2-1-3;;;;/h2*1-14H;1H2;2*1H;;/q;;;;;;+2/p-2

InChI key

CWHRHCATIKFSND-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Catalyst employed in the borylation of aryl halides with Bis(pinacolato)diboron(473294) as well as the coupling of the derived arylboronic esters with aryl halides leading to biaryls. Also used in the synthesis of lactams by CO insertion and medium-ring aryl ethers by intramolecular coupling of an aryl halide with an alcohol.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron, 62, 2006-2006 (2006)
Songnian Lin et al.
Journal of the American Chemical Society, 126(20), 6347-6355 (2004-05-20)
A full account of the total syntheses of proteasome inhibitors TMC-95A and -B is provided. A key feature of the syntheses involved installation of a cis-propenylamide moiety by a thermal rearrangement of an alpha-silylallyl amide. The scope and mechanism of
Bryan C Dickinson et al.
Nature protocols, 8(6), 1249-1259 (2013-06-01)
Mitochondria peroxy yellow 1 (MitoPY1) is a small-molecule fluorescent probe that selectively tracks to the mitochondria of live biological specimens and responds to local fluxes of hydrogen peroxide (H(2)O(2)) by a turn-on fluorescence enhancement. This bifunctional dye uses a triphenylphosphonium
Barry M Trost et al.
Organic letters, 6(11), 1745-1748 (2004-05-21)
An asymmetric synthesis of the benzazocine core of FR900482 has been achieved in 15 steps from 3,5-dinitro-p-toluic acid. Key features of the synthesis include an enantioselective N-methylephedrine-mediated zinc acetylide addition to a highly enolizable arylacetaldehyde and a novel Pd-catalyzed carbonylative
Tetrahedron, 62, 9002-9002 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service