Skip to Content
Merck
All Photos(1)

Documents

66294

Supelco

Methylcyclohexane

analytical standard

Synonym(s):

Hexahydrotoluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H11CH3
CAS Number:
Molecular Weight:
98.19
Beilstein:
505972
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

3.4 (vs air)

vapor pressure

37 mmHg ( 20 °C)
83.2 mmHg ( 37.7 °C)

Assay

≥99.5% (GC)

autoignition temp.

545 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

6.7 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.422 (lit.)
n20/D 1.422

bp

101 °C (lit.)

mp

−126 °C (lit.)

density

0.77 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

SMILES string

CC1CCCCC1

InChI

1S/C7H14/c1-7-5-3-2-4-6-7/h7H,2-6H2,1H3

InChI key

UAEPNZWRGJTJPN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methylcyclohexane [MCH] is a clear organic flammable liquid, mostly used as a solvent. During the development of chemical kinetic mechanisms for other cycloalkanes, MCH is considered as reaction starting point.

Application

It has been used in a study to develop low temperature oxidation mechanism, merging with high temperature mechanism which simulates MCH ignition in a rapid compression kinetic machine.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

24.8 °F - closed cup

Flash Point(C)

-4.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peter J. Wan, Phillip J. Wakelyn
Technology and Solvents for Extracting Oilseeds and Nonpetroleum Oils, 95-95 (1997)
Modeling and experimental investigation of methylcyclohexane ignition in a rapid compression machine.
Pitz, William J., et al.
Proceedings of the Combustion Institute, 31.1, 267-275 (2007)
Brian D Fitz et al.
Journal of chromatography. A, 1392, 82-90 (2015-03-31)
Low thermal mass gas chromatography (LTM-GC) was evaluated for rapid, high peak capacity separations with three injection methods: liquid, headspace solid phase micro-extraction (HS-SPME), and direct vapor. An Agilent LTM equipped with a short microbore capillary column was operated at

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service