Skip to Content
Merck
All Photos(2)

Key Documents

148938

Sigma-Aldrich

2-Iodopropane

contains copper as stabilizer, 99%

Synonym(s):

Isopropyl iodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHI
CAS Number:
Molecular Weight:
169.99
Beilstein:
1098244
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

99%

form

liquid

contains

copper as stabilizer

refractive index

n20/D 1.498 (lit.)

bp

88-90 °C (lit.)

mp

−90 °C (lit.)

density

1.703 g/mL at 25 °C (lit.)

functional group

alkyl halide
iodo

SMILES string

CC(C)I

InChI

1S/C3H7I/c1-3(2)4/h3H,1-2H3

InChI key

FMKOJHQHASLBPH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

2-Iodopropane is an organ iodine compound. It is commonly used as a homolytic and heterolytic alkylating agent and in the preparation of the α-isopropoxymethylene blocking group. Additionally, it acts as an intermediator in the conversion of thiocarbonate to alkene.[1]

Application

2-Iodopropane (Isopropyl iodide) was used to prepare butyric and isobutyric acid[2].

2-Iodopropane is used as an alkylating agent in the conversion of phenols, alcohols, oximes, carboxylic acids, and heteroatomic compounds (indoles, pyridines, purines, tetrazoles, pyrazoles, and amines) to give corresponding products.[1]

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

107.6 °F - closed cup

Flash Point(C)

42 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

2-Iodopropane
Wilson PD and Hilmey DG
Encyclopedia of Reagents for Organic Synthesis, Second Edition (2001)
Joe Rashan et al.
Journal of AOAC International, 86(4), 694-702 (2003-09-26)
An alternative liquid chromatographic (LC) method was developed and validated for the simultaneous determination of methoxyl and 2-hydroxypropoxyl substituents in hypromellose and hypromellose acetate succinate. The method uses the hydriodic acid cleavage reaction, catalyzed by adipic acid, of the substituted
Timur Coskun et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 19(21), 6840-6844 (2013-03-29)
Glycerol is converted to a mixture of butyric and isobutyric acid by rhodium- or iridium-catalysed carbonylation using HI as the co-catalyst. The initial reaction of glycerol with HI results in several intermediates that lead to isopropyl iodide, which upon carbonylation
W Kenealy et al.
Journal of bacteriology, 146(1), 133-140 (1981-04-01)
Iodopropane inhibited cell growth and methane production when Methanobacterium thermoautotrophicum, Methanobacterium formicicum, and Methanosarcina barkeri were cultured on H2-CO2. Iodopropane (40 microM) inhibited methanogenesis (30%) and growth (80%) when M. barkeri was cultured mixotrophically on H2-CO2-methanol. The addition of acetate
Development and validation of analytical procedure--control of residual 2-iodopropane in latanoprost.
Aleksandra Groman et al.
Acta poloniae pharmaceutica, 67(6), 673-676 (2011-01-15)

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service