Skip to Content
Merck
All Photos(3)

Key Documents

360066

Sigma-Aldrich

2,2,4-Trimethylpentane

anhydrous, 99.8%

Synonym(s):

TMP, Isooctane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2C(CH3)3
CAS Number:
Molecular Weight:
114.23
Beilstein:
1696876
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.21
grade:
anhydrous
Assay:
99.8%
technique(s):
UV/Vis spectroscopy: suitable
bp:
98-99 °C (lit.)
vapor pressure:
41 mmHg ( 21 °C)

grade

anhydrous

Quality Level

vapor density

3.9 (vs air)

vapor pressure

41 mmHg ( 21 °C)

Assay

99.8%

form

liquid

autoignition temp.

745 °F

expl. lim.

6 %

technique(s)

UV/Vis spectroscopy: suitable

impurities

<0.003% water
<0.005% water (100 mL pkg)

evapn. residue

<0.0003%

refractive index

n20/D 1.391 (lit.)

bp

98-99 °C (lit.)

mp

−107 °C (lit.)

density

0.692 g/mL at 25 °C (lit.)

SMILES string

CC(C)CC(C)(C)C

InChI

1S/C8H18/c1-7(2)6-8(3,4)5/h7H,6H2,1-5H3

InChI key

NHTMVDHEPJAVLT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,2,4-Trimethylpentane is a primary reference fuel with an arbitrary octane number of 100 as it burns smoothly without resulting in any knocking. It is mainly used to improve the octane rating of combustion fuels.

Application

2,2,4-Trimethylpentane has been used for the calibration of densimeter and for baseline correction in UV-Visible spectrophotometer.

Other Notes

The article number 360066-6X1L will be discontinued. Please order the single bottle 360066-1L which is physically identical with the same exact specifications.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

10.4 °F - closed cup

Flash Point(C)

-12 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Density, viscosity and specific heat capacity of diesel blends with rapeseed and soybean oil methyl ester
Albo PAG, et al.
Biomass and Bioenergy, 96, 87-95 (2017)
Confinement of DNA in water-in-oil microemulsions
Swami A, et al.
Langmuir, 24(20), 11828-11833 (2008)
Molecular structure of photosynthetic microbial biofuels for improved engine combustion and emissions characteristics
Hellier P, et al.
Frontiers in Bioengineering and Biotechnology, 3 (2015)
Nibedita Pal et al.
Analytical chemistry, 83(20), 7736-7744 (2011-09-09)
Fluorescence correlation spectroscopy (FCS) is an ideal tool for measuring molecular diffusion and size under extremely dilute conditions. However, the power of FCS has not been utilized to its best to measure diffusion and size parameters of complex chemical systems.
Emily E Fenn et al.
The Journal of chemical physics, 134(5), 054512-054512 (2011-02-10)
Water dynamics as reflected by the spectral diffusion of the water hydroxyl stretch were measured in w(0) = 2 (1.7 nm diameter) Aerosol-OT (AOT)/water reverse micelles in carbon tetrachloride and in isooctane solvents using ultrafast 2D IR vibrational echo spectroscopy.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service