Skip to Content
Merck
All Photos(2)

Key Documents

1.00789

Supelco

Ethyl acetate

for gas chromatography MS SupraSolv®

Synonym(s):

Essigester, Ethyl ester, EtOAc

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOC2H5
CAS Number:
Molecular Weight:
88.11
Beilstein:
506104
MDL number:
UNSPSC Code:
12191502
EC Index Number:
205-500-4
NACRES:
NA.03
Assay:
≥99.8% (GC)
technique(s):
GC/MS: suitable
bp:
76.5-77.5 °C (lit.)
vapor pressure:
73 mmHg ( 20 °C)
Pricing and availability is not currently available.

vapor density

3 (20 °C, vs air)

Quality Level

vapor pressure

73 mmHg ( 20 °C)

product line

SupraSolv®

Assay

≥99.8% (GC)

form

liquid

autoignition temp.

801 °F

potency

5620 mg/kg LD50, oral (Rat)
>18000 mg/kg LD50, skin (Rabbit)

expl. lim.

2.2-11.5 %, 38 °F

technique(s)

GC/MS: suitable

impurities

≤0.02% Water

evapn. residue

≤3.0 mg/L

color

APHA: ≤10

refractive index

n20/D 1.3720 (lit.)

bp

76.5-77.5 °C (lit.)

mp

−84 °C (lit.)

transition temp

flash point -4 °C

solubility

85.3 g/L

density

0.902 g/mL at 25 °C (lit.)

format

neat

storage temp.

2-30°C

SMILES string

CCOC(C)=O

InChI

1S/C4H8O2/c1-3-6-4(2)5/h3H2,1-2H3

InChI key

XEKOWRVHYACXOJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

SupraSolv MS is dedicated for use in gas chromatography coupled with mass spectrometric detection. SupraSolv solvents offer the largest specified retention time range, a clear baseline and a minimal signal-to-noise ratio leading to reliable and reproducible analysis results.

Application

  • Bio-guided isolation of a new sesquiterpene from Artemisia cina with anthelmintic activity against Haemonchus contortus L3 infective larvae.: This study explores the isolation of bioactive compounds from Artemisia cina, demonstrating the use of ethyl acetate in the extraction and purification processes. These findings may inspire chemists in academia to develop novel extraction techniques for natural products (Arango-De la Pava et al., 2024).
  • Attractant and repellent properties of Senna didymobotrya plant extracts to Amblyomma variegatum and Rhipicephalus appendiculatus.: This article details the use of ethyl acetate in extracting compounds from the Senna plant, which are tested for pesticidal properties, providing insights into its use in bioassay-guided fractionation (Kinyua et al., 2024).

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

SUPRASOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

24.8 °F

Flash Point(C)

-4 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Complete workflow for the comprehensive analysis of terpenes in cannabis

Complete workflow for the comprehensive analysis of terpenes in cannabis

Complete workflow for the comprehensive analysis of terpenes in cannabis

Complete workflow for the comprehensive analysis of terpenes in cannabis

Questions

  1. I don't see the type of packaging listed. Is this product available in a glass bottle?

    1 answer
    1. Yes, this material is packaged in glass bottles.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service