Skip to Content
Merck
All Photos(1)

Key Documents

68310

Supelco

2-Methylpentane

≥99.5% (GC), analytical standard, suitable for gas chromatography (GC) and HPLC

Synonym(s):

‘Isohexane’

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2CH(CH3)2
CAS Number:
Molecular Weight:
86.18
Beilstein:
1730735
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

Product Name

2-Methylpentane, analytical standard

grade

analytical standard

Quality Level

vapor density

3 (vs air)

vapor pressure

6.77 psi ( 37.7 °C)

Assay

≥99.5% (GC)

autoignition temp.

583 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

7 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.371 (lit.)
n20/D 1.371

bp

62 °C (lit.)

mp

−154 °C (lit.)

density

0.653 g/mL at 25 °C (lit.)

application(s)

petroleum

format

neat

SMILES string

CCCC(C)C

InChI

1S/C6H14/c1-4-5-6(2)3/h6H,4-5H2,1-3H3

InChI key

AFABGHUZZDYHJO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Methylpentane is a type of branched alkane classified under the volatile organic compounds (VOCs) family of chemicals.[1]

Application

2-Methylpentane has been used as a reference standard for the determination of 2-methylpentane as odor marker of lung cancer in human breath samples by gas chromatography-time of flight mass spectrometry (GC-TOF MS).[2] It may be used as a reference standard for the determination of 2-methylpentane in virgin olive oil samples by headspace solid-phase microextraction (SPME) followed by analysis using GC-MS coupled to flame ionization detector (FID),[3] in different microenvironments by multibed adsorption and short-path thermal desorption followed by GC-MS[4] and in exhaled air of patients with non-small cells lung cancer (NSCLC) by GC-MS.[5]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

19.4 °F - closed cup

Flash Point(C)

-7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification of volatile lung cancer markers by gas chromatography-mass spectrometry: comparison with discrimination by canines.
Buszewski B, et al.
Analytical and Bioanalytical Chemistry, 404(1), 141-146 (2012)
Exhaled volatile organic compounds in patients with non-small cell lung cancer: cross sectional and nested short-term follow-up study.
Diana P, et al.
Respiratory Research, 6(1), 71-71 (2005)
TD-GC-MS analysis of volatile metabolites of human lung cancer and normal cells in vitro.
Filipiak W, et al.
Cancer Epidemiology, Biomarkers & Prevention, 19(1), 182-195 (2010)
Determination of volatile organic compounds in different microenvironments by multibed adsorption and short-path thermal desorption followed by gas chromatographic-mass spectrometric analysis.
Kuntasal, et al.
Journal of Chromatography A, 1099(1-2), 43-54 (2005)
Analysis of virgin olive oil volatile compounds by headspace solid-phase microextraction coupled to gas chromatography with mass spectrometric and flame ionization detection.
Vichi, et al.
Journal of Chromatography A, 983(1-2), 19-33 (2003)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service