Skip to Content
Merck
All Photos(1)

Key Documents

D2773

Sigma-Aldrich

Dithranol

≥90% (HPLC)

Synonym(s):

1,8,9-Anthracenetriol, 1,8-Dihydroxy-9(10H)-anthracenone, 1,8-Dihydroxyanthrone, Anthralin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H10O3
CAS Number:
Molecular Weight:
226.23
Beilstein:
2054360
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

Assay

≥90% (HPLC)

form

solid

mp

178-181 °C (lit.)

solubility

chloroform: 20 mg/mL, clear to very slightly hazy, yellow

SMILES string

Oc1cccc2Cc3cccc(O)c3C(=O)c12

InChI

1S/C14H10O3/c15-10-5-1-3-8-7-9-4-2-6-11(16)13(9)14(17)12(8)10/h1-6,15-16H,7H2

InChI key

NUZWLKWWNNJHPT-UHFFFAOYSA-N

Gene Information

mouse ... Alox12(11684)

Looking for similar products? Visit Product Comparison Guide

General description

Dithranol (1, 8 - dihydroxy-9-anthrone) is also called as anthralin. It is a vital topical antipsoriatic drugs. Dithranol, an athracene derivative, is a free radical generating mitochondrial poison that induces apoptosis in keratinocytes.

Application

Dithranol, a potential anti-psoriasis medication, may be used to study its safety, efficacy, metabolism and pharmacological profile. Dithranol may be used to study is physicochemical properties, as a reference compound and to develop effective drug delivery vehicles. Dithranol may be used as a matrix for matrix assisted laser desorption/ionization imaging on a Fourier transform ion cyclotron resonance mass spectrometer.

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G C Hsieh et al.
Toxicology and applied pharmacology, 107(1), 16-26 (1991-01-01)
Primary cultured rat epidermal keratinocytes were used as an experimental model to detect oxidant-mediated adverse effects of dithranol (anthralin), a widely used antipsoriasis drug with tumor-promoting and skin-irritating properties. Keratinocytes were isolated and prepared from the skin of neonatal rats
Iyad Hailat et al.
Rapid communications in mass spectrometry : RCM, 28(2), 149-158 (2013-12-18)
Free sterols are neutral molecules that are difficult to analyze by MALDI or ESI and their molecular ions easily fragment. In order to increase their ionization efficiency and selectivity, sterols were derivatized by different reagents. Selected sterols were converted into
C Ronpirin et al.
Genetics and molecular research : GMR, 11(1), 412-420 (2012-03-01)
Although the precise causes of psoriasis remain to be elucidated, psoriasis has been known as a disorder in which factors in the immune system, enzymes and other biochemical substances that regulate skin cell division are functionally imbalanced, thereby resulting in
Wing Man Lau et al.
Pharmaceutics, 5(2), 232-245 (2013-12-05)
Psoriasis is a common, chronic and relapsing inflammatory skin disease. It affects approximately 2% of the western population and has no cure. Combination therapy for psoriasis often proves more efficacious and better tolerated than monotherapy with a single drug. Combination
Brian L Watson et al.
Physical chemistry chemical physics : PCCP, 17(24), 15788-15796 (2015-05-29)
A new sensitizer motif for dye sensitized solar cells (DSSC) has been developed. A heteroaromatic moiety containing a pyrazine ring links two porphyrin chromophores to the metal oxide surface via two carboxylic acid attachment groups. A test DSSC sensitized with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service