Skip to Content
Merck
All Photos(1)

Documents

37050

Supelco

Flupyradifurone

PESTANAL®, analytical standard

Synonym(s):

4-[(6-Chloro-3-pyridylmethyl)(2,2-difluoroethyl)amino]furan-2(5H)-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H11ClF2N2O2
CAS Number:
Molecular Weight:
288.68
Beilstein:
19518067
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

SMILES string

FC(F)CN(Cc1ccc(Cl)nc1)C2=CC(=O)OC2

InChI

1S/C12H11ClF2N2O2/c13-10-2-1-8(4-16-10)5-17(6-11(14)15)9-3-12(18)19-7-9/h1-4,11H,5-7H2

InChI key

QOIYTRGFOFZNKF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Flupyradifurone is a systemic insecticide, that belongs to the butenolide class of insecticides, which can be used for agricultural purposes in order to control a broad spectrum of sucking insects.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Target Organs

muscle

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicholas J Seiter et al.
Journal of economic entomology, 112(6), 2731-2736 (2019-09-11)
The sugarcane aphid, Melanaphis sacchari (Zehntner) (Hemiptera: Aphididae), has become a major pest of grain sorghum, Sorghum bicolor (L.) Moench, in the United States in recent years. Feeding by large densities of sugarcane aphids causes severe damage, which can lead
Paulo R R Barbosa et al.
Ecotoxicology (London, England), 26(5), 589-599 (2017-03-31)
Recent widespread infestations of the invasive sugarcane aphid, Melanaphis sacchari (Zehntner) (Hemiptera: Aphididae), in sorghum fields in the southern USA have created demand for insecticides that will provide effective control of sugarcane aphid, while conserving those beneficial species that contribute
An Evaluation of the Honey Bee (Hymenoptera: Apidae) Safety Profile of a New Systemic Insecticide, Flupyradifurone, Under Field Conditions in Florida
Campbell.WJ, et al.
Journal of Economic Entomology, 109(5), 1967-1972 (2016)
Priyadarshini Chakrabarti et al.
PloS one, 15(5), e0233033-e0233033 (2020-05-22)
Pesticide exposures can have detrimental impacts on bee pollinators, ranging from immediate mortality to sub-lethal impacts. Flupyradifurone is the active ingredient in Sivanto™ and sulfoxaflor is the active ingredient in Transform®. They are both relatively new insecticides developed with an
Flupyradifurone (Sivanto?) and its novel butenolide pharmacophore: Structural considerations
Jeschke P, et al.
Pesticide Biochemistry and Physiology, 121, 31-38 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service