Skip to Content
Merck
All Photos(3)

Key Documents

M5319

Sigma-Aldrich

Matrine

Synonym(s):

Matridin-15-one, Sophocarpidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H24N2O
CAS Number:
Molecular Weight:
248.36
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

2-8°C

SMILES string

O=C1CCCC2C3CCCN4CCCC(CN12)C34

InChI

1S/C15H24N2O/c18-14-7-1-6-13-12-5-3-9-16-8-2-4-11(15(12)16)10-17(13)14/h11-13,15H,1-10H2/t11-,12+,13+,15-/m0/s1

InChI key

ZSBXGIUJOOQZMP-JLNYLFASSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Matrine (MT) or Sophocarpidine is a naturally occurring bioactive compound, extracted from traditional Chinese herbs like Sophora flavescens and Radix Sophorae tonkinensis.[1]

Application

Matrine has been used:
  • to study its antioxidant and protective effect on doxorubicin (DOX)-induced cardiotoxicity in mice model and H9C2 cells[2]
  • as a chemotherapeutic agent to test its inhibitory effect on the growth of human glioblastoma multiforme (GBM) cells[3], and ovarian cancer cells[4]
  • to study its effect on mitochondrial fission and its anti-tumor functionality in HepG2 cells[5]
  • as a necroptosis inducer in cholangiocarcinoma (CCA) cell lines[6]

Biochem/physiol Actions

Matrine is a potent therapeutic agent with various pharmacological like anti-cancer, anti-bacterial, anti-fibrotic, anti-inflammatory properties.[1] It is a macropinocytosis inducer and decreases the cellular adenosine triphosphate (ATP) level.[7] Matrine may also serve as an effective sedative agent.[8] It serves as an anti-inflammatory agent in 2,4,6-trinitrobenzene sulfonic acid (TNBS)-induced intestinal inflammation in mice.[9] It functions to reverse the Warburg effect and suppress the growth of human colon cancer cells.[10]
Matrine is an alkaloid that is one of the major components in the root of the saphoro plant. Matrine has been studied for possible antiviral efficacy against hepatitis B and C, as well as impact against some skin diseases and forms of cancer.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arsenic trioxide-induced hERG K(+) channel deficiency can be rescued by matrine and oxymatrine through up-regulating transcription factor Sp1 expression.
Zhang Y, Dong Z, Jin L, et al.
Biochemical Pharmacology (2012)
Peiyuan Li et al.
Frontiers in physiology, 10, 28-28 (2019-02-26)
This study mainly investigated the effect of matrine on TNBS-induced intestinal inflammation in mice. TNBS treatment caused colonic injury and gut inflammation. Matrine (1, 5, and 10 mg/kg) treatment alleviated colonic injury and gut inflammation via reducing bleeding and diarrhea
Wenjing Zhou et al.
Cancer medicine, 7(9), 4729-4743 (2018-08-07)
Matrine, a traditional Chinese medicine, has recently been shown to have antitumor properties in diverse cancer cells. Here, we explored the effect of matrine on human glioblastoma multiforme (GBM) cells. Glioblastoma multiforme cell lines were treated with matrine to assess
Beibei Xu et al.
Cell death discovery, 3, 16096-16096 (2017-02-10)
The development of acquired resistance to pro-apoptotic antitumor agents is a major impediment to the cure of cholangiocarcinoma (CCA). Antitumor drugs inducing non-apoptotic cell death are considered as a new approach to overcome such drug resistance. Here, we reported for
Can Hu et al.
Acta pharmaceutica Sinica. B, 9(4), 690-701 (2019-08-07)
Oxidative stress and cardiomyocyte apoptosis are involved in the pathogenesis of doxorubicin (DOX)-induced cardiotoxicity. Matrine is well-known for its powerful anti-oxidant and anti-apoptotic capacities. Our present study aimed to investigate the effect of matrine on DOX-induced cardiotoxicity and try to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service