Skip to Content
Merck
All Photos(1)

Documents

31058

Supelco

λ-Cyhalothrin

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H19ClF3NO3
Molecular Weight:
449.85
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

SMILES string

CC1(C)[C@@H](\C=C(/Cl)C(F)(F)F)[C@H]1C(=O)O[C@H](C#N)c2cccc(Oc3ccccc3)c2

InChI

1S/C23H19ClF3NO3/c1-22(2)17(12-19(24)23(25,26)27)20(22)21(29)31-18(13-28)14-7-6-10-16(11-14)30-15-8-4-3-5-9-15/h3-12,17-18,20H,1-2H3/b19-12-/t17-,18+,20-/m0/s1

InChI key

ZXQYGBMAQZUVMI-GCMPRSNUSA-N

Looking for similar products? Visit Product Comparison Guide

General description

λ-Cyhalothrin is a type II highly active synthetic pyrethroid insecticide which contains a-cyano group.

Application

λ-Cyhalothrin is used as an internal standard for the determination of esfenvalerate from water by capillary gas chromatography with electron capture detection.
It may also be used as a pesticide reference standard for the determination of the analyte:
  • In virgin olive oil and cotton by gas chromatography (GC) with electron-capture detection.
  • In leafy vegetables by gas chromatography coupled with an ion trap mass spectrometry detection system (GC–ITMS).

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Development of a method for the determination of 9 currently used cotton pesticides by gas chromatography with electron capture detection.
Zhang B, et al.
Talanta, 75(4), 1055- 1060 (2008)
Determination of 23 pesticide residues in leafy vegetables using gas chromatography- ion trap mass spectrometry and analyte protectants.
Gonzalez-Rodriguez R, et al.
Journal of Chromatography A, 1196(4), 100-109 (2008)
Cypermethrin and lambda-cyhalothrin induced alterations in nucleic acids and protein contents in a freshwater fish, Channa punctatus.
Kumar A, Sharma B, Pandey RS.
Fish Physiology and Biochemistry, 34(4), 331-338 (2008)
Consequences of a short pulse of pesticide exposure for survival and reproduction of Gammarus pulex.
Cold A, Forbes VE.
Aquatic Toxicology (Amsterdam, Netherlands), 67(3), 287-299 (2004)
Determination of residues of endosulfan and five pyrethroid insecticides in virgin olive oil using gas chromatography with electron-capture detection.
Lentza-Rizos Ch, et al.
Journal of Chromatography A, 921(2), 297-304 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service