Skip to Content
Merck
All Photos(2)

Documents

32462

Supelco

Fluopyram

PESTANAL®, analytical standard

Synonym(s):

N-[2-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]ethyl]-2-(trifluoromethyl)benzamide, N-{2-[3-Chloro-5-(trifluoromethyl)-2-pyridyl]ethyl}-α,α,α-trifluoro-o-toluamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H11ClF6N2O
CAS Number:
Molecular Weight:
396.71
Beilstein:
11323270
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

FC(F)(F)c1cnc(CCNC(=O)c2ccccc2C(F)(F)F)c(Cl)c1

InChI

1S/C16H11ClF6N2O/c17-12-7-9(15(18,19)20)8-25-13(12)5-6-24-14(26)10-3-1-2-4-11(10)16(21,22)23/h1-4,7-8H,5-6H2,(H,24,26)

InChI key

KVDJTXBXMWJJEF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Fluopyram is a new pyridinylethylbenzamide fungicide. It is highly efficient in controlling a wide variety of pathogens such as Botrytis spp., Sclerotinia spp., and Monilia spp., and also against powdery mildews and some leaf spot diseases in a broad range of crops.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Fluopyram affects the fungal growth at all stages beginning from germination to sporulation. According to various studies, its biochemical mode of action is shown to rely on the inhibition of succinate dehydrogenase (complex II) that is present in the fungal mitochondrial respiratory chain, thus making it a succinate dehydrogenase inhibitor (SDHI).

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Zane J Grabau et al.
Journal of nematology, 52, 1-10 (2020-04-17)
Meloidogyne arenaria (peanut root-knot nematode (PRKN)) is a major pest of peanut. Nematicide application is an important tool for the management of PRKN. Nematicides with minimal effects on free-living nematodes are desired. Fluopyram nematicide is recently introduced in peanut production
Suchi Chawla et al.
Environmental science and pollution research international, 25(12), 11626-11634 (2018-02-13)
A field experiment was conducted to estimate residue persistence of fluopyram and its metabolite benzamide in cucumber fruits and soil and their risk assessment in humans and soil environment. Fluopyram (Kafka, 400 SC) was applied as soil drench twice at
Chao Xu et al.
Toxins, 11(5) (2019-05-18)
Deoxynivalenol (DON) is a class of mycotoxin produced in cereal crops infected with Fusarium graminearum species complex (FGSC). In China, FGSC mainly includes Fusarium asiaticum and F. graminearum. DON belongs to the trichothecenes and poses a serious threat to the
Response of microbial community to a new fungicide fluopyram in the silty-loam agricultural soil.
Zhang, Ying, et al.
Ecotox. Environ. Safety, 108, 273-280 (2014)
Wenyong Shao et al.
Molecular plant-microbe interactions : MPMI, 33(4), 580-589 (2020-01-11)
Succinate dehydrogenase (SDH) is an important respiratory enzyme which participates in the tricarboxylic acid cycle and oxidative phosphorylation. A previous study of the baseline sensitivity of Botrytis cinerea against SDH inhibitors (SDHIs) showed that intrinsic sensitivity of the small population

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service