Skip to Content
Merck
All Photos(1)

Key Documents

45368

Supelco

Carbendazim

PESTANAL®, analytical standard

Synonym(s):

BCM, Methyl 2-benzimidazolecarbamate, Methyl benzimidazol-2-ylcarbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H9N3O2
CAS Number:
Molecular Weight:
191.19
Beilstein:
649044
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

>300 °C (lit.)

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

COC(=O)Nc1nc2ccccc2[nH]1

InChI

1S/C9H9N3O2/c1-14-9(13)12-8-10-6-4-2-3-5-7(6)11-8/h2-5H,1H3,(H2,10,11,12,13)

InChI key

TWFZGCMQGLPBSX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Muta. 1B - Repr. 1B - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Perumal Rajakumar et al.
European journal of medicinal chemistry, 44(7), 3040-3045 (2008-09-10)
A series of carbazole based macrocyclic diamides with thia and oxy linkages have been synthesized and the inhibitory activity of the cyclophane amides against human pathogenic bacteria and plant pathogenic fungi are documented. (S)-1,10-Bi-2-naphthol [(S)-BINOL] based chiral carbazolophane amide emerged
Jie Lin et al.
Journal of natural products, 73(5), 905-910 (2010-04-23)
The new polyketides leptosphaerins A-G (1-7) and the known compounds monodictysin B (8), 2-(2,6-dihydroxybenzoyl)-3-hydroxy-5-methylbenzoic acid (9), and 2-(2,6-dihydroxy-4-methylbenzoyl)-6-hydroxybenzoic acid (10) have been isolated from solid cultures of the ascomycete fungus Leptosphaeria sp. The absolute configurations of compounds 1, 2, and
Qiong Chen et al.
European journal of medicinal chemistry, 43(3), 595-603 (2007-07-10)
In order to search novel agrochemicals with higher antifungal activity, a series of new 1,2,4-triazolo[1,5-a]pyrimidine derivatives bearing 1,3,4-oxadiazole moieties were designed and synthesized. Their antifungal activities against Rhizoctonia solani were evaluated in vitro. By determining the EC(50) values of all
D K Bayer et al.
Clinical and experimental immunology, 178(3), 459-469 (2014-07-22)
In areas without newborn screening for severe combined immunodeficiency (SCID), disease-defining infections may lead to diagnosis, and in some cases, may not be identified prior to the first year of life. We describe a female infant who presented with disseminated
Jae-Ran Lee et al.
Human mutation, 36(1), 69-78 (2014-09-30)
KIF1A is a neuron-specific motor protein that plays important roles in cargo transport along neurites. Recessive mutations in KIF1A were previously described in families with spastic paraparesis or sensory and autonomic neuropathy type-2. Here, we report 11 heterozygous de novo

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service