Skip to Content
Merck
All Photos(2)

Key Documents

840457C

Avanti

16:0-18:1 PG

Avanti Research - A Croda Brand

Synonym(s):

1-hexadecanoyl-2-(9Z-octadecenoyl)-sn-glycero-3-phospho-(1′racglycerol) (sodium salt); POPG; PG(16:0/18:1(9Z)); 110650

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H76O10PNa
CAS Number:
Molecular Weight:
770.99
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

description

1-palmitoyl-2-oleoyl-sn-glycero-3-phospho-(1′-rac-glycerol) (sodium salt), chloroform

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (840457C-25mg)
pkg of 2 × 20 mL (840457C-1g)
pkg of 2 × 4 mL (840457C-200mg)
pkg of 5 × 4 mL (840457C-500mg)

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

10 mg/mL (840457C-25mg)
25 mg/mL (840457C-1g)
25 mg/mL (840457C-200mg)
25 mg/mL (840457C-500mg)

lipid type

phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[Na+].[P](=O)([O-])(OC[C@H](OC(=O)CCCCCCC\C=C/CCCCCCCC)COC(=O)CCCCCCCCCCCCCCC)OCC(O)CO

InChI

1S/C40H77O10P.Na/c1-3-5-7-9-11-13-15-17-18-20-22-24-26-28-30-32-40(44)50-38(36-49-51(45,46)48-34-37(42)33-41)35-47-39(43)31-29-27-25-23-21-19-16-14-12-10-8-6-4-2;/h17-18,37-38,41-42H,3-16,19-36H2,1-2H3,(H,45,46);/q;+1/p-1/b18-17-;/t37?,38-;/m1./s1

InChI key

FJXDNGDRHUDFST-XQYKCTAGSA-M

General description

Phosphoglyceride (PG) are also referred as glycerophospholipid. It is an important component of membrane bilayers. It has three-carbon backbone of glycerol, two long-chain fatty acids, esterified to hydroxyl groups on carbons 1 and 2 of the glycerol and phosphoric acid esterified to the C3 hydroxyl group of glycerol. It is found at high levels in the membranes of all cells and at low levels in fat stores. 16:0-18:1 PG/1-palmitoyl-2-oleoyl-sn-glycero-3-phospho-(1′-rac-glycerol) (POPG) is an anionic lipid, which possesses one negative charge per lipid.

Application

16:0-18:1 PG has been used in liposome preparation. It is suitable for use as a component in monoolein to treat purified β2-adrenergic receptor (β2AR)-T4 lysozyme (T4L)- Gαs α5 helix (GsCT)-CC chimera protein and in lipid vesicle preparation.

Biochem/physiol Actions

Phosphoglycerides (PG)/glycerophospholipids play a key role in cell signaling systems. It also serves as an anchor for proteins in cell membranes.

Packaging

30 mL Amber Narrow Mouth Glass Bottle with Screw Cap (840457C-1g)
5 mL Clear Glass Sealed Ampule (840457C-200mg)
5 mL Clear Glass Sealed Ampule (840457C-25mg)
5 mL Clear Glass Sealed Ampule (840457C-500mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structural insights into the process of GPCR-G protein complex formation
Liu X, et al.
Cell, 177(5), 1243-1251 (2019)
Ionization Properties of Phospholipids Determined by Zeta Potential Measurements
Sathappa M and Alder NN
Bio-protocol, 6(22) (2016)
Membrane Structure and Dynamics
Cell Biology, 227-239 (2017)
Lipids
Medical Biochemistry, 99-119 (2017)
Mechanism of four de novo designed antimicrobial peptides
Murray B, et al.
Test, 291(49), 25706- 25715 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service