Skip to Content
Merck
All Photos(1)

Key Documents

100600C

Avanti

E. coli Extract Polar

Avanti Research - A Croda Brand

Synonym(s):

E. coli Polar Lipid Extract

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

chloroform solution

packaging

pkg of 2 × 20 mL (100600C-1g)
pkg of 1 × 4 mL (100600C-100mg)
pkg of 5 × 4 mL (100600C-500mg)

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

25 mg/mL (100600C-100mg)
25 mg/mL (100600C-1g)
25 mg/mL (100600C-500mg)

lipid type

lipid extracts

shipped in

dry ice

storage temp.

−20°C

General description

This product is an extract of E. coli B (ATCC 11303) grown in Kornberg Minimal media at 37°C and taken at 3/4 log growth phase. Total E. coli lipid extract is a chloroform:methanol extract of the respective tissue. This extract is partitioned against deionized water and the chloroform phase is concentrated. Polar lipid extract is the total lipid extract precipitated with acetone and then extracted with diethyl ether.

Application

E. coli Extract Polar is suitable to prepare small unilamellar vesicles, to mimic the surface of the E. coli inner membrane.
E. coli extract polar has been for the preparation of liposomes.
E. coli Extract Polar has been used:
  • in unilamellar liposomes preparation for kinetic studies
  • in the reconstitution of ATP-binding cassette (ABC) transporter for electron paramagnetic resonance spectroscopy studies
  • in the reconstitution of molybdate transport protein (MolBC) for continuous wave-electron paramagnetic resonance studies

Biochem/physiol Actions

E.coli Polar lipid extracts comprise phosphatidylethanolamine as major lipid component. The minor components include the phosphatidylglycerol (PG) and cardiolipin. The leaflets of cytoplasmic membrane and inner leaflet of outer membrane are the source of E. coli lipid extract. It is used for reconstituting membrane proteins and forms bilayer by vesicle fusion.

Packaging

30 mL Amber Narrow Mouth Glass Bottle with Screw Cap (100600C-1g)
5 mL Clear Glass Sealed Ampule (100600C-100mg)
5 mL Clear Glass Sealed Ampule (100600C-500mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The
Enzymology Primer for Recombinant DNA Technology (2017)
Small substrate transport and mechanism of a molybdate ATP binding cassette transporter in a lipid environment
Rice AJ, et al.
The Journal of biological chemistry, 289(21), 15005-15013 (2014)
An inducible reconstitution system for the real-time kinetic analysis of protease activity and inhibition inside the membrane
Baker RP and Urban S
Methods in Enzymology, 584, 229-253 (2017)
Emily Peterson et al.
Biochimica et biophysica acta, 1808(2), 516-521 (2010-10-26)
Amantadine-sensitive proton uptake by liposomes is currently the preferred method of demonstrating M2 functionality after reconstitution, to validate structural determination with techniques such as solid-state NMR. With strong driving forces (two decades each of both [K(+)] gradient-induced membrane potential and
Sushant Kumar et al.
The Journal of biological chemistry, 295(1), 55-68 (2019-11-09)
Single-chain antibodies from camelids have served as powerful tools ranging from diagnostics and therapeutics to crystallization chaperones meant to study protein structure and function. In this study, we isolated a single-chain antibody from an Indian dromedary camel (ICab) immunized against

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service