Skip to Content
Merck
All Photos(2)

Key Documents

840455P

Avanti

16:0 PG

Avanti Research - A Croda Brand

Synonym(s):

1,2-dihexadecanoyl-sn-glycero-3-phospho-(1′racglycerol) (sodium salt); DPPG; PG(16:0/16:0), 1,2-dipalmitoyl-sn-glycero-3-phospho-(1′-rac-glycerol) (sodium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H74O10PNa
CAS Number:
Molecular Weight:
744.95
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 g (840455P-1g)
pkg of 1 × 200 mg (840455P-200mg)
pkg of 1 × 25 mg (840455P-25mg)
pkg of 1 × 500 mg (840455P-500mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[Na+].[P](=O)([O-])(OC[C@H](OC(=O)CCCCCCCCCCCCCCC)COC(=O)CCCCCCCCCCCCCCC)OCC(O)CO

InChI

1S/C38H75O10P.Na/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-37(41)45-33-36(34-47-49(43,44)46-32-35(40)31-39)48-38(42)30-28-26-24-22-20-18-16-14-12-10-8-6-4-2;/h35-36,39-40H,3-34H2,1-2H3,(H,43,44);/q;+1/p-1/t35?,36-;/m1./s1

InChI key

LDWIWSHBGAIIMV-ODZMYOIVSA-M

General description

Phosphoglyceride (PG) are also referred as glycerophospholipid. It is an important component of membrane bilayers. It has three-carbon backbone of glycerol, two long-chain fatty acids, esterified to hydroxyl groups on carbons 1 and 2 of the glycerol and phosphoric acid esterified to the C3 hydroxyl group of glycerol. It is found at high levels in the membranes of all cells and at low levels in fat stores.

Application

16:0 PG has been used as a component in internal lipid standard mixture for matrix-assisted laser desorption/ionization (MALDI)- fourier-transform ion cyclotron resonance (FTICR)- mass spectrometry (MS) analysis. It is also suitable to use in liposome flotation assay to study the specificity of protein-lipid interaction.

Biochem/physiol Actions

Phosphoglycerides (PG)/glycerophospholipids play a key role in cell signaling systems. It also serves as an anchor for proteins in cell membranes.

Packaging

20 mL Clear Glass Screw Cap Vial (840455P-1g)
20 mL Clear Glass Screw Cap Vial (840455P-500mg)
5 mL Amber Glass Screw Cap Vial (840455P-200mg)
5 mL Amber Glass Screw Cap Vial (840455P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ridwan P Putra et al.
Langmuir : the ACS journal of surfaces and colloids, 35(50), 16576-16582 (2019-11-26)
Protein-encapsulated nanoclusters (NCs) are emerging as a versatile platform for in-vivo imaging and other biomedical applications due to their ultrasmall size and excitation in the near-infrared region. Encapsulation may however affect protein structure, size, charge, and its interaction with lipid
Membrane Structure and Dynamics
Cell Biology, 227-239 (2017)
Lipids
Medical Biochemistry, 99-119 (2017)
Carolina Medina Amado et al.
Biochimica et biophysica acta. Biomembranes, 1862(2), 183135-183135 (2019-11-19)
The mechanism of action of the anti-Listeria peptide enterocin CRL35 was studied with biophysical tools by using lipid mixtures that mimicked Gram-positive plasma membranes. Langmuir monolayers and infrared spectroscopy indicated that the peptide readily interacted with phospholipid assembled in monolayers
Changes in lipid distribution in E. coli strains in response to norfloxacin
Schenk ER, et al.
Journal of Mass Spectrometry : Jms, 50(1), 88-94 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service