Skip to Content
Merck
All Photos(3)

Key Documents

860062C

Avanti

Brain SM

Avanti Research - A Croda Brand

Synonym(s):

18:0 SM; Octadecanoyl Sphingomyelin; N-octadecanoyl-D-erythro-sphingosylphosphorylcholine; N-(octadecanoyl)-sphing-4-enine-1-phosphocholine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

description

Sphingomyelin (Brain, Porcine), chloroform

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (860062C-25mg)
pkg of 2 × 4 mL (860062C-200mg)

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

10 mg/mL (860062C-25mg)
25 mg/mL (860062C-200mg)

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[P](=O)([O-])(OC[C@H](NC(=O)CCCCCCCCCCCCCCCCC)[C@H](O)\C=C\CCCCCCCCCCCCC)OCC[N+](C)(C)C

InChI

1S/C41H83N2O6P/c1-6-8-10-12-14-16-18-20-21-23-25-27-29-31-33-35-41(45)42-39(38-49-50(46,47)48-37-36-43(3,4)5)40(44)34-32-30-28-26-24-22-19-17-15-13-11-9-7-2/h32,34,39-40,44H,6-31,33,35-38H2,1-5H3,(H-,42,45,46,47)/b34-32+/t39-,40+/m0/s1

InChI key

LKQLRGMMMAHREN-YJFXYUILSA-N

General description

As a major constituent of cell membranes, sphingomyelin is found at particularly high concentrations in the membranes of nerve cells (in the myelin sheaths) and red blood cells. It was previously thought to have a purely structural role, similar to the function of phosphatidylcholine, through intermolecular interactions mediated by the 2-amide group, the 3-hydroxy group and the 4,5-trans double bond of the sphingoid base. However, it is now appreciated that sphingomyelin has a high affinity for cholesterol and that these two lipids pack tightly into liquid-ordered domains among a liquid-disordered phase to form lipid rafts. These membrane microdomains are thought to function as signaling platforms that regulate the localization and interactions of proteins. But sphingomyelin does not just influence signaling as a component of lipid rafts — it is also a precursor to ceramides and other sphingolipid metabolites that comprise the sphingomyelin cycle or sphingolipid network.
Sphingomyelin and related lipids

Application

Brain SM may be used:
  • for ion-mobility spectrometry–mass spectrometry (IMS-MS)
  • for lipid mixture in giant unilamellar vesicles
  • as a growth medium for madin-darby canine kidney (MDCK) cells

Packaging

5 mL Clear Glass Sealed Ampule (860062C-200mg)
5 mL Clear Glass Sealed Ampule (860062C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Opposing kinesin and myosin-I motors drive membrane deformation and tubulation along engineered cytoskeletal networks
McIntosh B B,et al.
Current Biology, 28(2), 236-248 (2018)
Inhibition of acid sphingomyelinase depletes cellular phosphatidylserine and mislocalizes K-Ras from the plasma membrane
Cho K J,et al.
Molecular and cellular biology, 36(2), 363-374 (2016)
Yusuke Sato et al.
Acta biomaterialia, 102, 341-350 (2019-11-17)
Despite the fact that small-sized lipid nanoparticles (LNPs) are important for improved tissue penetration and efficient drug delivery, their poor stability and intracellular trafficking significantly hinders their use as potent small-sized LNPs. It has been reported that both the diffusion
Tomokazu Yasuda et al.
Langmuir : the ACS journal of surfaces and colloids, 34(44), 13426-13437 (2018-10-24)
In this study, we applied fluorescence spectroscopy, differential scanning calorimetry (DSC), and 2H NMR to elucidate the properties of nanoscopic segregated domains in stearoylsphingomyelin (SSM)/dioleoylphosphatidylcholine (DOPC) and dihydrostearoylsphingomyelin (dhSSM)/DOPC binary membranes. The results obtained from fluorescence measurements suggest the existence
Sphingomyelin metabolism at the plasma membrane: implications for bioactive sphingolipids.
Milhas D, et al.
Febs Letters, 584, 1887-1894 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service