Skip to Content
Merck
All Photos(2)

Documents

437247

Sigma-Aldrich

Poly(ethylene-co-vinyl acetate)

vinyl acetate 12 wt. %, melt index 8 g/10 min (190°C/2.16kg)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH2CH2)m[CH2CH(OCOCH3)]n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

beads

autoignition temp.

500 °F

melt index

8 g/10 min (190°C/2.16kg)

contains

400-1200 ppm BHT as antioxidant

composition

vinyl acetate, 12 wt. %

hardness

43 (Shore D, ASTM D 2240)

mp

95 °C

transition temp

softening point 65 °C (Vicat, ASTM D 1525)

density

0.933 g/mL at 25 °C

SMILES string

C=C.CC(=O)OC=C

InChI

1S/C4H6O2.C2H4/c1-2-3-4(5)6;1-2/h2H,1,3H2,(H,5,6);1-2H2

InChI key

DQXBYHZEEUGOBF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Flexible tubing, color concentrates, gaskets and molded parts for autos, plastic lenses and pumps.

Features and Benefits

Thermoplastic, resilient, tough and resistant to ozone and environmental stress cracking. Inherently flexible, requiring no plasticizers.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Neide Pena Coto et al.
Dental traumatology : official publication of International Association for Dental Traumatology, 28(2), 108-113 (2011-07-28)
There has been a significant increase in the number of facial fractures stemming from sport activities in recent years, with the nasal bone one of the most affected structures. Researchers recommend the use of a nose protector, but there is
Taiji Ogawa et al.
International journal of oral science, 4(1), 14-18 (2012-03-17)
Bacterial infection associated with the use of medical or dental devices is a serious concern. Although devices made of ethylene vinyl acetate (EVA) are often used in the oral cavity, there are no established standards for their storage. We investigated
Ethylene vinyl acetate (foam): an inexpensive and useful tool for teaching suture techniques in dermatologic surgery.
Daniela Gutiérrez-Mendoza et al.
Dermatologic surgery : official publication for American Society for Dermatologic Surgery [et al.], 37(9), 1353-1357 (2011-05-28)
Rafael Denadai et al.
Acta cirurgica brasileira, 27(3), 271-278 (2012-03-31)
To assess the acquisition of suture skills by training on ethylene-vinyl acetate bench model in novice medical students. Sixteen medical students without previous surgery experience (novices) were randomly divided into two groups. During one hour group A trained sutures on
Shijun Sheng et al.
Journal of separation science, 36(4), 721-728 (2013-01-29)
In this report, a graphene/poly(ethylene-co-vinyl acetate) composite electrode was fabricated by melt compounding for the amperometric detection of capillary electrophoresis. The composite electrode was fabricated by packing a mixture of graphene and melted poly(ethylene-co-vinyl acetate) in a piece of fused

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service