Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Poly(ethylene-
co
-vinyl acetate)
Linear Formula:
(CH
2
CH
2
)
m
[CH
2
CH(OCOCH
3
)]
n
CAS Number:
24937-78-8
Products
Brand
Sigma-Aldrich (1)
Melting Point (°C)
Physical Form
Chemical Composition
Polymer Composition
Polymer Type
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
340502
Poly(ethylene-
co
-vinyl acetate),
vinyl acetate 40 wt. %, melt index (41-63 dg/min (190°C/2.16kg)), contains 190-910 ppm inhibitor
Expand
View Pricing
437247
Poly(ethylene-
co
-vinyl acetate),
vinyl acetate 12 wt. %, melt index 8 g/10 min (190°C/2.16kg)
Expand
View Pricing
437239
Poly(ethylene-
co
-vinyl acetate),
vinyl acetate 18 wt. %, melt index 8 g/10 min (190°C/2.16kg), contains 200-900 ppm BHT as inhibitor
Expand
View Pricing