Skip to Content
Merck
All Photos(2)

Key Documents

426717

Sigma-Aldrich

Poly(ethylene-co-acrylic acid)

acrylic acid 5 wt. %, beads

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH2CH2)x[CH2CH(CO2H)]y
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

description

drop point 105 °C Mettler

form

beads

composition

acrylic acid, 5 wt. %

hardness

≤2.5 dmm (ASTM D 5, 25 °C)

viscosity

575 cP(145 °C, Brookfield)(lit.)

acid number

37‑44 mg KOH/g

transition temp

Tm 99-101 °C

density

0.93 g/mL at 25 °C

SMILES string

C=C.OC(=O)C=C

InChI

1S/C3H4O2.C2H4/c1-2-3(4)5;1-2/h2H,1H2,(H,4,5);1-2H2

InChI key

QHZOMAXECYYXGP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Processing and performance additive. Promotes crystallization of PET. Assists dispersion of additives in plastics.

Features and Benefits

Forms reversible ionic clusters (crosslinks). Promotes adhesion to various substrates, tougher, more chemically resistant and more transparent than parent acid copolymer.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ning Luo et al.
Colloids and surfaces. B, Biointerfaces, 50(2), 89-96 (2006-06-06)
Poly(ethylene-co-acrylic acid) (EAA) films were reacted with glycine, 12-aminododecanoic acid, aspartic acid, 5-aminoisophthalic acid, ethanolamine, diethylamine, dimethylamine, N-isopropylamine, and dimethylaminoethyleneamine to prepare EAA films with negatively charged, non-charged, hydrophilic, and hydrophobic functionalities. Attenuated total reflectance Fourier transform infrared spectroscopy, differential
Newton C Fawcett et al.
Langmuir : the ACS journal of surfaces and colloids, 20(16), 6651-6657 (2004-07-28)
Yoshimoto et al. [Anal. Chem. 2002, 74, 4306-4309] reported that a quartz crystal microbalance or QCM changed its response to sucrose solutions according to its angle of immersion. The effect was tentatively attributed to gravity-caused stress on the viscous interface
A P Martínez-Camacho et al.
Carbohydrate polymers, 91(2), 666-674 (2012-11-06)
The obtaining of chitosan extruded films was possible by using low density polyethylene (LDPE) as a matrix polymer and ethylene-acrylic acid copolymer as an adhesive, in order to ensure adhesion in the interphase of the immiscible polymers. The obtained blend
Optician's occupational allergic contact dermatitis, paresthesia and paronychia caused by anaerobic acrylic sealants.
L Kanerva et al.
Contact dermatitis, 44(2), 117-119 (2001-02-24)
Marc David Grynbaum et al.
Journal of chromatography. A, 1156(1-2), 80-86 (2006-12-01)
The increased demand for chromatographic materials that are able to achieve a fast separation of large quantities of structure analogues is a great challenge. It is known that polymer based chromatographic materials have a higher loadability, compared to silica based

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service