Skip to Content
Merck
All Photos(3)

Key Documents

P2443

Sigma-Aldrich

Pluronic® F-127

powder, BioReagent, suitable for cell culture

Synonym(s):

Non-ionic copolymer surfactant

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C3H6O·C2H4O)x
CAS Number:
MDL number:
UNSPSC Code:
12161902
PubChem Substance ID:
NACRES:
NA.75

description

Non-ionic
contains 100ppm BHT

product line

BioReagent

form

powder

mol wt

~12600 g/mol

technique(s)

cell culture | mammalian: suitable

CMC

950-1000 ppm (~25°C)

transition temp

cloud point >100 °C

HLB

18 - 23

SMILES string

O2C(C2)C.O1CC1

InChI

1S/C3H6O.C2H4O/c1-3-2-4-3;1-2-3-1/h3H,2H2,1H3;1-2H2

InChI key

RVGRUAULSDPKGF-UHFFFAOYSA-N

General description

Pluronicpluronic® F-127, also known as poloxamer 407, is a non-toxic and nonionic copolymer. It contains polyethylene oxide-polypropylene oxide arranged in a triblock structure to form PEO-PPO-PEO symmetric. Surfactant properties of this polymer derived from its amphiphilic nature, allow them to interact with hydrophobic surfaces and biological membranes.

Application

Pluronic® F-127 has been used:

  • as a delivery system for a peptide to the wound site to study the effect of the peptide on wound healing in mice.
  • as a surface coating for PDMS (polydimethylsiloxane) stamps to make them hydrogel repellent.
  • in the preparation of calcium indicator stock solution for measuring neural activities.
  • as a component of phosphate-buffered saline (PBS), to lower unspecific cell and protein adhesion to a PDMS-based microfluidic device.
  • This non-ionic copolymer surfactant is suitable for use in insect cell culture applications as an antifoaming agent.

Biochem/physiol Actions

Pluronic® F-127 is a thermo-reversible gel that finds its application in different industries due to its high thermosensitivity, biodegradability, and biocompatibility properties. These properties make this polymer suitable for use as a versatile drug carrier.Pluronic® F-127 improves the therapeutic efficiency of drugs by increasing their bioavailability and thermostability. Additionally, it can promote cellular attachment and collagen formation, which facilitates angiogenesis and tissue regeneration. Due to its efficacy, Pluronic F-127 has been utilized in the encapsulation of mesenchymal stem cells (MSCs) to aid in the regeneration of tissues with low levels of vascularity, such as epithelial tissues, adipose tissues, cartilage, tendons, and bone tissues.

Legal Information

Pluronic is a registered trademark of BASF

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dual-responsive (pH/temperature) Pluronic F-127 hydrogel drug delivery system for textile-based transdermal therapy
Chatterjee S, et al.
Scientific Reports, 9(1) (2019)
Environmentally responsive injectable materials
Bearat HH and Vernon BL
Academic Emergency Medicine : Official Journal of the Society for Academic Emergency Medicine, 263-297 (2011)
Connexin43 carboxyl-terminal peptides reduce scar progenitor and promote regenerative healing following skin wounding
Ghatnekar GS, et al.
Regenerative Medicine, 4(2), 205-223 (2009)
Federico Ceriani et al.
Proceedings of the National Academy of Sciences of the United States of America, 113(46), E7194-E7201 (2016-11-04)
Spatially and temporally coordinated variations of the cytosolic free calcium concentration ([Ca2+]c) play a crucial role in a variety of tissues. In the developing sensory epithelium of the mammalian cochlea, elevation of extracellular adenosine trisphosphate concentration ([ATP]e) triggers [Ca2+]c oscillations
NeuroCa: integrated framework for systematic analysis of spatiotemporal neuronal activity patterns from large-scale optical recording data
Jang MJ and Nam Y
Neurophotonics, 2(3) (2015)

Articles

Local delivery of bioactive molecules using an implantable device can decrease the amount of drug dose required as well as non-target site toxicities compared to oral or systemic drug administration.

Related Content

Tissue engineering fabricates tissues cultures from scaffolds, living cells, and biologically active molecules by simulating the microenvironment of the body to repair or replace damaged tissue.

Tissue engineering fabricates tissues cultures from scaffolds, living cells, and biologically active molecules by simulating the microenvironment of the body to repair or replace damaged tissue.

Tissue engineering fabricates tissues cultures from scaffolds, living cells, and biologically active molecules by simulating the microenvironment of the body to repair or replace damaged tissue.

Tissue engineering fabricates tissues cultures from scaffolds, living cells, and biologically active molecules by simulating the microenvironment of the body to repair or replace damaged tissue.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service