Skip to Content
Merck
All Photos(2)

Documents

432121

Sigma-Aldrich

Ytterbium(III) fluoride

anhydrous, powder, 99.98% trace metals basis

Synonym(s):

Ytterbium trifluoride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
YbF3
CAS Number:
Molecular Weight:
230.04
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

grade

anhydrous

Assay

99.98% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: ytterbium

impurities

≤250.0 ppm Trace Rare Earth Analysis

density

~8.20 g/mL at 25 °C (lit.)

SMILES string

F[Yb](F)F

InChI

1S/3FH.Yb/h3*1H;/q;;;+3/p-3

InChI key

XASAPYQVQBKMIN-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Employed in the preparation and study of fluorinated glasses.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J. Mater. Sci., 31, 2981-2981 (1996)
Svetlana N Pleskova et al.
Micron (Oxford, England : 1993), 90, 23-32 (2016-08-28)
Nanoparticles are increasingly being used for treatment and diagnostic purposes, but their effects on cells is not fully understood. Here, the interaction of fluorescent up-conversion nanoparticles (UpC-NPs) with neutrophils was investigated by imaging and measurement of membrane-cytosceletal elasticity by atomic
Michele Vano et al.
American journal of dentistry, 22(6), 366-370 (2010-02-25)
To evaluate the effects of storage condition and duration on the resistance to fracture of different fiber post systems (and to morphologically assess the post structure before and after storage. Three types of fiber posts (DT Light Post, GC Post
Anders Lindberg et al.
Acta odontologica Scandinavica, 62(2), 97-101 (2004-06-17)
This in vitro study evaluated curing depths of a universal hybrid resin composite with two viscosities (Tetric Ceram and Tetric Flow) after curing with 6 different quartz tungsten halogen and light-emitting diode curing units. Irradiance (light intensity) of the curing
J Rud et al.
Journal of endodontics, 22(9), 477-480 (1996-09-01)
A retrograde root-end cover with a special resin composite (Retroplast) combined with the dentin bonding agent Gluma (Bayer AG) has been used since 1984 by the authors. Its content of silver, added to promote radiopacity, has been found to lower

Articles

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service