Merck
  • Optimization of supercritical fluid consecutive extractions of fatty acids and polyphenols from Vitis vinifera grape wastes.
Optimization of supercritical fluid consecutive extractions of fatty acids and polyphenols from Vitis vinifera grape wastes.
Journal of food science (2014-12-05)
Oier Aizpurua-Olaizola, Markel Ormazabal, Asier Vallejo, Maitane Olivares, Patricia Navarro, Nestor Etxebarria, Aresatz Usobiaga
ABSTRACT
In this study, supercritical fluid extraction has been successfully applied to a sequential fractionation of fatty acids and polyphenols from wine wastes (2 different vitis vinifera grapes). To this aim, in a 1st step just fatty acids were extracted and in a 2nd one the polyphenols. The variables that affected to the extraction efficiency were separately optimized in both steps following an experimental design approach. The effect of extraction temperature flow, pressure, and time were thoroughly evaluated for the extraction of fatty acids, whereas the addition of methanol was also considered in the case of the polyphenols extraction. A quantitative extraction with high efficiency was achieved at a very short time and low temperatures. Concerning quantification, fatty acids were determined by means of gas chromatography coupled to mass spectrometry after a derivatization step, whereas the polyphenols were analyzed by means of high performance liquid chromatography coupled to tandem mass spectrometry and the Folin-Ciocalteu method.
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Methyl decanoate, ≥99%, FG
Sigma-Aldrich
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Supelco
Methyl elaidate, analytical standard
Supelco
Methyl decanoate, analytical standard
Supelco
Methyl myristoleate, analytical standard
Sigma-Aldrich
Methyl myristate, ≥98%, FG
Sigma-Aldrich
Sodium carbonate, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Methyl stearate, ≥96%, FG
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sulfuric acid, 99.999%
Supelco
Methyl myristate, analytical standard
Supelco
Sulfuric acid, for the determination of nitrogen, ≥97.0%
Supelco
Sodium carbonate, reference material for titrimetry, certified by BAM, >99.5%
Supelco
Methyl palmitate, analytical standard
Supelco
Methyl palmitoleate, analytical standard
Supelco
Methyl linoleate, analytical standard
Sigma-Aldrich
Methyl decanoate, 99%
Sigma-Aldrich
Methyl palmitate, ≥97%
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Methyl elaidate, ≥99% (capillary GC)
Sigma-Aldrich
Methyl arachidate, ≥99% (capillary GC)
Sigma-Aldrich
Methyl stearate, ~99% (GC)
Sigma-Aldrich
Methyl palmitoleate, ≥99% (capillary GC), liquid
Sigma-Aldrich
Methyl palmitate, ≥99% (capillary GC)
Sigma-Aldrich
Methyl linoleate, ≥98% (GC)
Sigma-Aldrich
Methyl myristate, ≥99% (GC)
Sigma-Aldrich
Methyl tridecanoate, ≥97% (GC)
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Methyl myristoleate, ≥98.5% (capillary GC), liquid