Skip to Content
Merck
All Photos(1)

Key Documents

W272205

Sigma-Aldrich

Methyl myristate

≥98%, FG

Synonym(s):

Methyl tetradecanoate, Myristic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)12COOCH3
CAS Number:
Molecular Weight:
242.40
FEMA Number:
2722
Beilstein:
1773739
EC Number:
Council of Europe no.:
387
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.106
NACRES:
NA.21
Organoleptic:
fatty; waxy
grade:
FG
Kosher
biological source:
synthetic
Agency:
meets purity specifications of JECFA
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 172.515

Assay

≥98%

refractive index

n20/D 1.436 (lit.)

bp

323 °C (lit.)

mp

18 °C (lit.)

density

0.855 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fatty; waxy

SMILES string

CCCCCCCCCCCCCC(=O)OC

InChI

1S/C15H30O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15(16)17-2/h3-14H2,1-2H3

InChI key

ZAZKJZBWRNNLDS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl myristate is a fatty acid methyl ester commonly used as a flavoring essence.
It is one of the main:
  • phytochemical constituent of Cycas Beddomei cones
  • volatile flavor component of γ-aminobutyric acid (GABA) tea

Application


  • Modulating the hydrophobicity of cellulose by lipase-catalyzed transesterification.: This research discusses the use of methyl myristate for modifying cellulose properties through enzymatic processes, which can be crucial for developing hydrophobic materials for various industrial applications (Sharma et al., 2024).

  • Biosynthesis of insect sex pheromone precursors via engineered β-oxidation in yeast.: Demonstrates the synthesis of methyl myristate in yeast as a precursor to pheromones, suggesting its potential in bioengineering for sustainable agricultural practices (Petkevicius et al., 2022).

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

233.6 °F - closed cup

Flash Point(C)

112.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

GC-MS determination of bioactive constituents of Cycas beddomei cones.
Kumar NR, et al.
International Journal of Pharmacy and Biological Sciences, 3(3), 344-350 (2012)
Study on flavour volatiles of ?-aminobutyric acid (GABA) green tea.
Zeng Z, et al.
African Journal of Biotechnology, 11(51), 11333-11341 (2012)
Enzymatic synthesis of methyl myristate using thermocol-immobilized lipase.
Sharma CK & Kanwar SS.
International Journal of Recent Scientific Research, 3(5), 407-412 (2012)
Saeedeh Noushini et al.
Scientific reports, 10(1), 19799-19799 (2020-11-15)
Diverse methods have been used to sample insect semiochemicals. Sampling methods can differ in efficiency and affinity and this can introduce significant biases when interpreting biological patterns. We compare common methods used to sample tephritid fruit fly rectal gland volatiles
Balasubramanian Narasimhan et al.
Bioorganic & medicinal chemistry letters, 16(11), 3023-3029 (2006-03-24)
A series of esters and amides of myristic acid was synthesized and tested in vitro for antibacterial activity against gram-positive and gram-negative bacteria. All the compounds showed activity comparable to that of the standard drug, ciprofloxacin. The structural characteristics governing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service