Skip to Content
Merck
All Photos(1)

Key Documents

L1876

Sigma-Aldrich

Methyl linoleate

≥98% (GC)

Synonym(s):

Linoleic acid methyl ester, Methyl cis,cis-9,12-octadecadienoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3(CH2CH=CH)2(CH2)7CO2CH3
CAS Number:
Molecular Weight:
294.47
Beilstein:
1727614
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥98% (GC)

form

liquid

refractive index

n20/D 1.462 (lit.)

bp

192 °C/4 mmHg (lit.)

mp

−35 °C (lit.)

density

0.889 g/mL at 25 °C (lit.)

functional group

ester

lipid type

omega FAs

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCC\C=C/C\C=C/CCCCCCCC(=O)OC

InChI

1S/C19H34O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19(20)21-2/h7-8,10-11H,3-6,9,12-18H2,1-2H3/b8-7-,11-10-

InChI key

WTTJVINHCBCLGX-NQLNTKRDSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl linoleate, non-conjugated, is more easily oxidized than conjugated methyl linoleate .

Application

The oxidation of methyl linoleate in organic solution and phosphatidylcholine (PC) in liposomal membranes induced by an azo initiator has often been used as a model for lipid peroxidation in vivo .

Biochem/physiol Actions

Methyl linoleate may be used as a model compound in oxidation/peroxidation assays to evaluate the anti-peroxidation activity of fullerenes, ellagitannins and other natural products.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marja Kähkönen et al.
Journal of agricultural and food chemistry, 60(5), 1167-1174 (2012-01-11)
Ellagitannins from red raspberries (Rubus idaeus) and cloudberries (Rubus chamaemorus) were isolated by using column chromatography and preparative HPLC. The berry phenolic isolates consisted of 80% (cloudberry) and of 60% (raspberry) of ellagitannins, with raspberries also containing anthocyanins. The main
Fanny Bery et al.
International journal of molecular sciences, 21(13) (2020-07-10)
Hypoxia is a well-established feature of prostate cancer (PCa) and is associated with disease aggressiveness. The hypoxic microenvironment initiates multiple adaptive responses including epithelial-to-mesenchymal transition (EMT) and a remodeling of calcium homeostasis involved in cancer progression. In the present study
Steven Lim et al.
Journal of hazardous materials, 390, 121532-121532 (2019-12-18)
In this research, biomass from oil palm empty fruit bunch was used as the carbon precursor and sulfonated by 4-benzenediazonium sulfonate (4-BDS) to produce solid acid catalyst. The as-synthesized catalysts were characterized and the performances were tested in esterification of
Guo-Xiang Li et al.
European journal of medicinal chemistry, 45(5), 1821-1827 (2010-02-04)
5,7-, 5,8-, 6,8-, 7,8-dichloro-4-quinolinol-3-carboxylic acid (5,7-, 5,8-, 6,8-, 7,8-DCQA) together with 7-chloro-4-quinolinol-3-carboxylic acid (7-CQA) and 4-quinolinol-3-carboxylic acid (QA) were synthesized to investigate the antioxidant properties. 5,7-DCQA exhibited the highest ability to scavenge 2,2'-azinobis(3-ethylbenzothiazoline-6-sulfonate) cationic radical (ABTS+.), 2,2'-diphenyl-1-picrylhydrazyl (DPPH) and galvinoxyl
Roman Shchepin et al.
Journal of the American Chemical Society, 132(49), 17490-17500 (2010-11-26)
Free radical co-oxidation of polyunsaturated lipids with tyrosine or phenolic analogues of tyrosine gave rise to lipid peroxide-tyrosine (phenol) adducts in both aqueous micellar and organic solutions. The novel adducts were isolated and characterized by 1D and 2D NMR spectroscopy

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service