Skip to Content
Merck
All Photos(1)

Key Documents

45119

Supelco

Methyl elaidate

analytical standard

Synonym(s):

Elaidic acid methyl ester, Methyl trans-9-octadecenoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7CH=CH(CH2)7COOCH3
CAS Number:
Molecular Weight:
296.49
Beilstein:
1727038
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.450

mp

9-10 °C

density

0.871 g/mL at 20 °C (lit.)

format

neat

functional group

ester

shipped in

ambient

storage temp.

2-8°C

SMILES string

CCCCCCCC\C=C\CCCCCCCC(=O)OC

InChI

1S/C19H36O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19(20)21-2/h10-11H,3-9,12-18H2,1-2H3/b11-10+

InChI key

QYDYPVFESGNLHU-ZHACJKMWSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Byung-Jun Kollbe Ahn et al.
Journal of agricultural and food chemistry, 60(9), 2179-2189 (2012-02-07)
Toxic solvent and strong acid catalysts causing environmental issues have been mainly used for ring-opening of epoxidized oleochemicals. Here, we demonstrated that magnesium stearate (Mg-stearate) was a high efficient catalyst for solvent-free ring-opening of epoxidized methyl oleate, a model compound
Renee M Thomas et al.
Journal of the American Chemical Society, 133(19), 7490-7496 (2011-04-23)
N-Aryl,N-alkyl N-heterocyclic carbene (NHC) ruthenium metathesis catalysts are highly selective toward the ethenolysis of methyl oleate, giving selectivity as high as 95% for the kinetic ethenolysis products over the thermodynamic self-metathesis products. The examples described herein represent some of the
Farshad Darvishi et al.
New biotechnology, 28(6), 756-760 (2011-02-18)
The yeast Yarrowia lipolytica degrades efficiently low-cost hydrophobic substrates for the production of various added-value products such as lipases. To obtain yeast strains producing high levels of extracellular lipase, Y. lipolytica DSM3286 was subjected to mutation using ethyl methanesulfonate (EMS)
Atanu Biswas et al.
Journal of agricultural and food chemistry, 56(14), 5611-5616 (2008-06-19)
An environmentally friendly water-based pathway to form the azide derivatives of soybean oil and fatty esters is reported. This entails first the formation of epoxides and then the azidization of the epoxides. The azidization reaction is carried out at high
Ursula Biermann et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 18(26), 8201-8207 (2012-05-18)
A kinetic study of the dodecanethiol-catalyzed cis/trans isomerization of methyl oleate (cis-2) without added initiator was performed by focusing on the initiation of the radical chain reaction. The reaction orders of the rate of isomerization were 2 and 0.5 for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service