W505501
Methyl decanoate
≥99%, FG
Synonym(s):
Capric acid methyl ester, Decanoic acid methyl ester, Methyl caprate
About This Item
Fragrance grade
Halal
Recommended Products
biological source
synthetic
grade
FG
Fragrance grade
Halal
Agency
follows IFRA guidelines
reg. compliance
EU Regulation 1223/2009
EU Regulation 1334/2008 & 872/2012
FDA 21 CFR 110
Assay
≥99%
refractive index
n20/D 1.425 (lit.)
bp
108 °C/10 mmHg
224 °C (lit.)
mp
−14-−11 °C (lit.)
density
0.871 g/mL at 20 °C (lit.)
application(s)
flavors and fragrances
Documentation
see Safety & Documentation for available documents
food allergen
no known allergens
fragrance allergen
no known allergens
Organoleptic
oily; fruity; wine-like
SMILES string
CCCCCCCCCC(=O)OC
InChI
1S/C11H22O2/c1-3-4-5-6-7-8-9-10-11(12)13-2/h3-10H2,1-2H3
InChI key
YRHYCMZPEVDGFQ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 2
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
230.9 °F - Pensky-Martens closed cup
Flash Point(C)
110.5 °C - Pensky-Martens closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Protocols
HPLC Analysis of Fatty Acid Methyl Esters (FAMES) on SUPELCOSIL™ LC-18
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service