Merck
  • Negative selection by apoptosis enriches progenitors in naïve and expanded human umbilical cord blood grafts.
Negative selection by apoptosis enriches progenitors in naïve and expanded human umbilical cord blood grafts.
Bone marrow transplantation (2014-04-30)
K Mizrahi, S Ash, T Peled, I Yaniv, J Stein, N Askenasy
ABSTRACT
The influence of TNF-α and Fas-ligand (FasL) on viability and function was evaluated in fresh- and expanded-umbilical cord blood (UCB) cells. CD34(+) progenitors and T cells display outstanding survival, whereas ~30% and >50% B lymphocytes and myeloid cells undergo spontaneous apoptosis within 24 and 48 h, respectively. Although the impact of exposure to toxic doses of FasL and TNF-α was undetectable in measurements of apoptosis; removal of dead cells after 2 days of incubation with the ligands revealed a twofold increase in frequency of colony-forming cells (CFU). The sensitivity of progenitors to apoptosis was also unaffected by Fas cross-linking following TNF-induced upregulation of the receptor, increasing CFU frequency without impairing SCID repopulating cell (SRC) activity. Most significant enrichment in CD34(+) progenitors and corresponding increase in CFU frequency were observed when FasL was applied during the final week of ex vivo expansion under the influence of nicotinamide, without impairing SRC activity. These data emphasize differential sensitivities of UCB progenitors and lineage-positive cells to apoptotic signaling mediated by the Fas and TNF receptors, which might be useful in improving the efficiency of ex vivo expansion and improving UCB cell engraftment.
Sigma-Aldrich
5-Carboxy-fluorescein diacetate N-succinimidyl ester, for fluorescence, ≥95.0% (HPLC)
Sigma-Aldrich
5(6)-Carboxyfluorescein diacetate N-succinimidyl ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Tumor Necrosis Factor-α from mouse, TNF-α, recombinant, expressed in E. coli, powder, suitable for cell culture
Sigma-Aldrich
MISSION® esiRNA, targeting human FAS
Sigma-Aldrich
TNF-α human, Animal-component free, recombinant, expressed in E. coli, suitable for cell culture
Sigma-Aldrich
Human Fas Ligand ELISA Kit, for serum, plasma, and cell culture supernatants
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Nicotinamide, ≥99.5% (HPLC)
Sigma-Aldrich
Nicotinamide, ≥98.5% (HPLC)
Sigma-Aldrich
DL-Tyrosine, 99%
Sigma-Aldrich
Tumor Necrosis Factor-α from rat, TNF-α, recombinant, expressed in E. coli, powder, suitable for cell culture
Sigma-Aldrich
Tumor Necrosis Factor-α human, TNF-α, recombinant, expressed in E. coli, powder, suitable for cell culture
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Niacinamide, meets USP testing specifications
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
SAFC
L-Glutamine
Sigma-Aldrich
Nicotinamide, ≥98% (HPLC), powder
Sigma-Aldrich
Nicotinamide, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Glutamine
Supelco
Niacinamide, Pharmaceutical Secondary Standard; Certified Reference Material
Nicotinamide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Tumor Necrosis Factor-α human, Xeno-free, recombinant, expressed in HEK 293 cells, suitable for cell culture
Sigma-Aldrich
Human Fas ELISA Kit, for cell and tissue lysates
Sigma-Aldrich
Human Fas ELISA Kit, for serum, plasma, cell culture supernatant and urine
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Niacinamide, United States Pharmacopeia (USP) Reference Standard
Tyrosine, European Pharmacopoeia (EP) Reference Standard