Skip to Content
Merck
All Photos(2)

Key Documents

N5535

Sigma-Aldrich

Niacinamide

meets USP testing specifications

Synonym(s):

Nicotinamide, Niacinamide, Nicotinic acid amide, Pyridine-3-carboxylic acid amide, Vitamin B3, Vitamin PP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6N2O
CAS Number:
Molecular Weight:
122.12
Beilstein:
383619
EC Number:
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.21

biological source

synthetic (organic)

Quality Level

Agency

USP/NF
meets USP testing specifications

Assay

98.5-101.5% dry basis

form

crystals

color

white

mp

128-131 °C (lit.)

application(s)

pharmaceutical (small molecule)

SMILES string

NC(=O)c1cccnc1

InChI

1S/C6H6N2O/c7-6(9)5-2-1-3-8-4-5/h1-4H,(H2,7,9)

InChI key

DFPAKSUCGFBDDF-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Niacinamide, also known as 3-pyridinecarboxamide, is a water soluble vitamin. It is widely present in plants, animal food and cosmetics.

Application

Niacinamide has been used:
  • as a component of islet maturation culture media
  • to induce differentiation of insulin secreting cells
  • to compare its preclinical effectiveness with that of progesterone

Biochem/physiol Actions

Niacinamide is a primary precursor of nicotinamide adenine dinucleotide (NAD+), which is an important coenzyme in adenosine triphosphate (ATP) production. Nicotinamide deficiency is associated with pellagra.
Nicotinamide is an amide derivative of vitamin B3 and a PARP inhibitor

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

302.0 °F - closed cup

Flash Point(C)

150 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Necrostatin-1 supplementation enhances young porcine islet maturation and in vitro function
Lau H, et al.
Xenotransplantation (2019)
Role of nicotinamide in DNA damage, mutagenesis, and DNA repair
Surjana D, et al.
Journal of Nucleic Acids, 2010 (2010)
Stimulatory Role of Glycated Fetal Bovine Serum Along with Iron on In Vitro Production of Insulin by Differentiated Mouse Embryonic Stem Cells
Nishigaki I, et al.
Journal of Health Science, 57(4), 356-361 (2011)
A comparison of the effects of nicotinamide and progesterone on functional recovery of cognitive behavior following cortical contusion injury in the rat
Peterson TC, et al.
Journal of Neurotrauma, 29(18), 2823-2830 (2012)
Robert J Motzer et al.
The Lancet. Oncology, 14(6), 552-562 (2013-04-20)
In a phase 3 trial comparing the efficacy and safety of axitinib versus sorafenib as second-line treatment for metastatic renal cell carcinoma, patients given axitinib had a longer progression-free survival (PFS). Here, we report overall survival and updated efficacy, quality

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service