Skip to Content
Merck

Skip To

21888

5(6)-Carboxyfluorescein diacetate N-succinimidyl ester

BioReagent, suitable for fluorescence, ≥90% (HPLC)

Synonym(s):

CFSE

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C29H19NO11
CAS Number:
Molecular Weight:
557.46
NACRES:
NA.32
PubChem Substance ID:
UNSPSC Code:
12352108
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

suitable as fluorogenic substrate for esterases

Quality Segment

product line

BioReagent

assay

≥90% (HPLC)

form

solid

mol wt

557.46 g/mol

color

almost white to yellow

mp

152-154 °C (lit.)

solubility

DMF: soluble, DMSO: soluble

fluorescence

λex 492 nm; λem 517 nm in 0.1 M Tris pH 8.0 (esterase)

suitability

suitable for fluorescence

storage temp.

−20°C

SMILES string

CC(=O)Oc1ccc2c(Oc3cc(OC(C)=O)ccc3C24OC(=O)c5cc(ccc45)C(=O)ON6C(=O)CCC6=O)c1.CC(=O)Oc7ccc8c(Oc9cc(OC(C)=O)ccc9C8%10OC(=O)c%11ccc(cc%10%11)C(=O)ON%12C(=O)CCC%12=O)c7

InChI

1S/2C29H19NO11/c1-14(31)37-17-4-7-21-23(12-17)39-24-13-18(38-15(2)32)5-8-22(24)29(21)20-6-3-16(11-19(20)28(36)40-29)27(35)41-30-25(33)9-10-26(30)34;1-14(31)37-17-4-7-20-23(12-17)39-24-13-18(38-15(2)32)5-8-21(24)29(20)22-11-16(3-6-19(22)28(36)40-29)27(35)41-30-25(33)9-10-26(30)34/h2*3-8,11-13H,9-10H2,1-2H3

InChI key

LZFDYQHOQDFMOA-UHFFFAOYSA-N

General description

5-(and-6)-Carboxyfluorescein diacetate N-succinimidyl ester (CFSE) is a fluorescent dye, which is membrane permeable. It labels cells by covalently binding to intracellular molecules. CFSE is used to track lymphocyte migration and proliferation.

Application

5(6)-Carboxyfluorescein diacetate N-succinimidyl ester (CFSE) is an amine reactive fluorescent probe used to stain cells, especially immune cells, for analysis in applications such as flow cytometry. Forms green fluorescent conjugates on deacetylation.
Amine-reactive label for long term tracing of cells. Forms green fluorescent conjugates on deacetylation.
5-Carboxyfluorescein diacetate N-succinimidyl ester has been used:
  • to label cells for flow cytometry
  • in lymphocyte proliferation assay
  • to determine the intracellular or intra-organellar pH in chloroplasts
  • to label Leishmania amazonensis and Leishmania major

Biochem/physiol Actions

5(6)-Carboxyfluorescein diacetate N-succinimidyl ester is used as an indicator for esterase activity.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
218842187921878
description

BioReagent, suitable for fluorescence, ≥90% (HPLC)

description

BioReagent, suitable for fluorescence, ≥85% (HPCE)

description

BioReagent, suitable for fluorescence, ≥90.0% (HPLC)

description

BioReagent, suitable for fluorescence, mixture of isomers, ≥80% (HPLC)

suitability

suitable for fluorescence

suitability

suitable for fluorescence

suitability

suitable for fluorescence

suitability

suitable for fluorescence

fluorescence

λex 492 nm; λem 517 nm in 0.1 M Tris pH 8.0 (esterase)

fluorescence

λex 470 nm; λem 529 nm in 0.1 M Tris pH 8.0 (esterase)

fluorescence

λex 492 nm; λem 517 nm in 0.1 M Tris pH 8.0 (esterase)

fluorescence

λex 492 nm; λem 517 nm in DMF

color

almost white to yellow

color

-

color

-

color

-

solubility

DMF: soluble, DMSO: soluble

solubility

methanol: soluble, DMSO: soluble, DMF: soluble

solubility

DMSO: soluble

solubility

DMF: soluble, DMSO: soluble

form

solid

form

-

form

powder

form

powder

assay

≥90% (HPLC)

assay

≥85% (HPCE)

assay

≥90.0% (HPLC)

assay

≥80% (HPLC)


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–7 of 7 Questions  
  1. Hi. I bought your CFSE. I want to ask how to use it? I follow one paper: doi: 10.1016/j.xcrm.2025.102027. They used CFSE in the final concentration of 0.5 uM for 3 mins at RT. Is this you suggested? And if yes, how to prepare 0.5 uM?

    1 answer
    1. No direct reconstitution protocol is available for this product. Based on an online literature search, the CFSE dye, purchased as carboxyfluorescein diacetate succinimidyl ester (MW 557.47 g/mole), is typically provided in 25 mg vials. To prepare a final stock solution of 5 mM, dissolve the 25 mg in 8.96 mL DMSO. This solution can be stored as 50-100 μL aliquots at -20 °C for several months. It is important to avoid exposure to aqueous solutions during storage. Note that the CFSE dye is at a final concentration of 5 μM, however, the optimum concentration may have to be determined for each batch prepared, and then checked every 6 months to ensure it is effective.

      Helpful?

  2. What is the in vivo (mouse) half-life of CFDA-SE? What is the route of clearance (kidney, liver?)

    1 answer
    1. The half-life is not determined for this product.
      Please see the link below to a publication that may be helpful:
      https://www.semanticscholar.org/paper/The-Use-of-Carboxyfluorescein-Diacetate-Ester-to-Quah-Parish/cae3435e063cb9f46cea4ddfc7c560f5821e19b3

      Please note that, unless otherwise stated, this product is intended for research use only and is not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

      Helpful?

  3. Is Product 21888, 5(6)-Carboxyfluorescein diacetate N-succinimidyl ester, fluorescent? Are conjugates of this compound fluorescent?

    1 answer
    1. No. This compound and conjugates of this compound become fluorescent only when acted upon by an enzyme such as esterase. The enzyme removes the acetate groups.

      Helpful?

  4. Is Product 21888, 5(6)-Carboxyfluorescein diacetate N-succinimidyl ester, light sensitive?

    1 answer
    1. Yes. The powder and solutions (especially solutions) should be kept protected from ambient light whenever possible.

      Helpful?

  5. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  6. What is the ratio between the (5) and (6) isomer of Product 21888, 5(6)-Carboxyfluorescein diacetate N-succinimidyl ester?

    1 answer
    1. Our Quality Control group does not routinely report the ratio; however, it is very likely that both isomers are present.

      Helpful?

  7. At what concentration is Product 21888, 5(6)-Carboxyfluorescein diacetate N-succinimidyl ester, soluble in DMSO?

    1 answer
    1. We have found 5(6)-Carboxyfluorescein diacetate N-succinimidyl ester to be soluble in DMSO at a concentration of 1 mg/mL.

      Helpful?

Reviews

No rating value

Active Filters