Skip to Content
Merck

Skip To

G6392

L-Glutamine

γ-irradiated, BioXtra, suitable for cell culture

Synonym(s):

(S)-2,5-Diamino-5-oxopentanoic acid, L-Glutamic acid 5-amide, Levoglutamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
H2NCOCH2CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
146.14
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
EC Number:
200-292-1
NACRES:
NA.75
Beilstein/REAXYS Number:
1723797
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

sterility

γ-irradiated

product line

BioXtra

form

powder

packaging

vial of 0.292 g

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

color

white to faint yellow

mp

185 °C (dec.) (lit.)

solubility

H2O: 10 mL/vial, clear to slightly hazy

SMILES string

N[C@@H](CCC(N)=O)C(O)=O

InChI

1S/C5H10N2O3/c6-3(5(9)10)1-2-4(7)8/h3H,1-2,6H2,(H2,7,8)(H,9,10)/t3-/m0/s1

InChI key

ZDXPYRJPNDTMRX-VKHMYHEASA-N

Gene Information

rat ... Ggt1(116568)

Application

L-Glutamine has been used as a media supplement:
  • for cochlear organotypic culturing
  • in Iscove′s modified Dulbecco′s medium for HeLa cells culturing
  • in Dulbecco′s modified Eagle′s medium for primary hepatocyte cell culturing

Qualified for use in cell culture applications, especially serum-free cell culture.

Biochem/physiol Actions

Glutamine mediates protein turnover via cellular mTOR (mammalian target of rapamycin) signalling. It is also known to be associated with the inhibition of apoptosis.
L-Glutamine is an essential amino acid that is a crucial component of culture media that serves as a major energy source for cells in culture. L-Glutamine is very stable as a dry powder and as a frozen solution. In liquid media or stock solutions, however, L-glutamine degrades relatively rapidly. Optimal cell performance usually requires supplementation of the media with L-glutamine prior to use.
L-glutamine is an essential amino acid that is a crucial component of culture media, and serves as a major energy source for cells in culture. Optimal cell performance usually requires supplementation of the medium with L-glutamine prior to use.

Packaging

50 mL amber serum vial with rubber stopper.

Preparation Note

Prepares a 200 mM solution when reconstituted to 10 mL with sterile water.

Other Notes

The dipeptide L-Ala-LGln is frequently used in modern serum-free media formulations because it provides a more stable form of glutamine in vitro.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
G8540765231.00286
form

powder

form

powder

form

-

form

solid

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

API processing | cocrystal formation: suitable, API processing | salt formation: suitable

product line

BioXtra

product line

-

product line

TraceCERT®

product line

EMPROVE® EXPERT

impurities

endotoxin, tested

impurities

endotoxin, residual solvents, tested

impurities

-

impurities

-

mp

185 °C (dec.) (lit.)

mp

185 °C (dec.) (lit.)

mp

185 °C (dec.) (lit.)

mp

185 °C (dec.) (lit.)

solubility

H2O: 10 mL/vial, clear to slightly hazy

solubility

H2O: 25 mg/mL

solubility

-

solubility

100 g/L


Still not finding the right product?

Explore all of our products under L-Glutamine


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–2 of 2 Questions  
  1. Is the molecular weight of L-Glutamine 146.14 or 146.15? The label information changed.

    1 answer
    1. Based on a molecular formula of C5H10N2O3, the molecular weight to seven significant figures is 146.1456. Over the years, this value may have been reported in Sigma-Aldrich documentation or on labels as either 146.15 (by rounding up) or 146.14 (by truncation). The material itself has not changed.

      Helpful?

  2. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

Reviews

No rating value

Active Filters