Skip to Content
Merck
All Photos(6)

Key Documents

G3126

Sigma-Aldrich

L-Glutamine

≥99% (HPLC), ReagentPlus®

Synonym(s):

(S)-2,5-Diamino-5-oxopentanoic acid, L-Glutamic acid 5-amide, Levoglutamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NCOCH2CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
146.14
Beilstein:
1723797
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26
Pricing and availability is not currently available.

Product Name

L-Glutamine, ReagentPlus®, ≥99% (HPLC)

Quality Level

product line

ReagentPlus®

Assay

≥99% (HPLC)

form

powder

impurities

≤0.2% free ammonia.
~0.5% Glutamic acid

color

white to off-white

mp

185 °C (dec.) (lit.)

solubility

1 M HCl: 50 mg/mL, clear to slightly hazy, colorless

cation traces

<0.2% (Ammonia (NH3))

application(s)

cell analysis
peptide synthesis

functional group

amide

storage temp.

room temp

SMILES string

N[C@@H](CCC(N)=O)C(O)=O

InChI

1S/C5H10N2O3/c6-3(5(9)10)1-2-4(7)8/h3H,1-2,6H2,(H2,7,8)(H,9,10)/t3-/m0/s1

InChI key

ZDXPYRJPNDTMRX-VKHMYHEASA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

L-Glutamine is a versatile amino acid that plays an essential role in various metabolic processes within the human body. This non-essential amino acid is naturally abundant and contributes significantly to several vital functions. One of its primary functions is acting as the primary carrier of nitrogen in the body. It serves as the transport vehicle for nitrogen, which is a fundamental building block for many biological processes. Additionally, L-Glutamine is a major energy source for numerous types of cells, ensuring they have the energy they need to function optimally.

Moreover, L-Glutamine is essential for maintaining a delicate balance of ammonia in the body. This balance is crucial for preventing toxic ammonia levels. This amino acid facilitates the conversion of glutamic acid to glutamine, a process central to ammonia regulation. L-Glutamine is not only involved in nitrogen transport and energy production but also serves as a key player in various metabolic pathways. It participates in the biosynthesis of proteins, similar to other amino acids. Furthermore, it plays a significant role in the synthesis of urea and purines, vital components for nucleic acid production. L-Glutamine is also a vital resource for generating cellular energy, working closely with glucose for this purpose.

Another essential function of L-Glutamine is its contribution to nitrogen donation for multiple anabolic processes. These processes are vital for building crucial molecules, including purines, which are crucial for DNA production. This amino acid also provides carbon, replenishing the citric acid cycle, a central process in energy production within cells. L-Glutamine is a critical amino acid, acting as a nitrogen carrier, energy source, and contributing to protein synthesis and ammonia regulation. Its versatile functions make it a molecule of significant interest in the fields of cell biology, biochemical research, and metabolomics investigations.

Application

L-Glutamine has been used to study the effects of amino acids in promoting food consumption in Drosophila melanogaster.[1]It has also been used to study non-enzymatic gluconeogenesis.[2]

Biochem/physiol Actions

L-glutamine is the most abundant amino acid in the body. It is essential for the synthesis of L-asparagine.[3][4] L-glutamic acid aids the incorporation of NH4+ into biomolecules.[5]
L-Glutamine is an essential amino acid that is a crucial component of culture media that serves as a major energy source for cells in culture. L-Glutamine is very stable as a dry powder and as a frozen solution. In liquid media or stock solutions, however, L-glutamine degrades relatively rapidly. Optimal cell performance usually requires supplementation of the media with L-glutamine prior to use.

Features and Benefits

  • Can be used in Metabolomics and Biochemical research
  • High-quality compound suitable for multiple research applications

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Natural Health Dictionary: Your comprehensive A-to Z guide for healing with herbs, nutrition, supplements, and secret remedies (2011)
Advances in Food and Nutrition Research, 58-58 (1917)
An internal sensor detects dietary amino acids and promotes food consumption in Drosophila
Zhe Yang
Nature, 1-36 (2017)
Lehninger Principles of Biochemistry Edition 4th (2005)
Nonenzymatic gluconeogenesis-like formation of fructose 1,6-bisphosphate in ice.
Messner CB
Proceedings of the National Academy of Sciences of the USA, 114(28), 7403-7407 (2017)

Questions

1–4 of 4 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  4. Is the molecular weight of L-Glutamine 146.14 or 146.15? The label information changed.

    1 answer
    1. Based on a molecular formula of C5H10N2O3, the molecular weight to seven significant figures is 146.1456. Over the years, this value may have been reported in Sigma-Aldrich documentation or on labels as either 146.15 (by rounding up) or 146.14 (by truncation). The material itself has not changed.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service