Skip to Content
Merck
All Photos(1)

Key Documents

Z3541

Sigma-Aldrich

trans-Zeatin-riboside

suitable for plant cell culture, BioReagent, ~95%

Synonym(s):

9-(β-D-Ribofuranosyl)-trans-zeatin, N6-(trans-4-Hydroxy-3-methyl-2-buten-1-yl)adenosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H21N5O5
CAS Number:
Molecular Weight:
351.36
Beilstein:
5460894
MDL number:
UNSPSC Code:
10171502
PubChem Substance ID:
NACRES:
NA.72

product line

BioReagent

Assay

~95%

technique(s)

cell culture | plant: suitable

application(s)

agriculture

storage temp.

−20°C

SMILES string

C\C(CO)=C/CNc1ncnc2n(cnc12)[C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O

InChI

1S/C15H21N5O5/c1-8(4-21)2-3-16-13-10-14(18-6-17-13)20(7-19-10)15-12(24)11(23)9(5-22)25-15/h2,6-7,9,11-12,15,21-24H,3-5H2,1H3,(H,16,17,18)/b8-2+/t9-,11-,12-,15-/m1/s1

InChI key

GOSWTRUMMSCNCW-HNNGNKQASA-N

Looking for similar products? Visit Product Comparison Guide

Application

Trans-Zeatin-riboside has been used to treat the crystals, retrieved from the capillaries for crystallization. It has also been used as a supplement in Murashige and Skoog (MS) medium to form shoot induction medium.

Biochem/physiol Actions

Trans-zeatin-riboside is a kind of cytokinin precursor, which is an important long-distance signalling form in xylem vessels. It serves as an inhibitory factor in root xylem sap by negatively modulating adventitious root formation.
Koenig RL; Morris RO; Polacco JC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Systemic transport of trans-zeatin and its precursor have differing roles in Arabidopsis shoots
Osugi A, et al.
Nature plants, 3(8), 17112-17112 (2017)
An efficient marker-free vector for clean gene transfer into plants
Xin C and Guo J
African Journal of Biotechnology, 11(16), 3751-3757 (2012)
A trans-zeatin riboside in root xylem sap negatively regulates adventitious root formation on cucumber hypocotyls
Kuroha T, et al.
Journal of Experimental Botany, 53(378), 2193-2200 (2002)
Structure of mistletoe lectin I from Viscum album in complex with the phytohormone zeatin
Meyer A, et al.
Biochimica et Biophysica Acta (BBA)-Proteins and Proteomics, 1784(11), 1590-1595 (2008)
Jianchang Yang et al.
Planta, 228(1), 137-149 (2008-03-15)
Early-flowered superior spikelets usually exhibit a faster grain filling rate and heavier grain weight than late-flowered inferior spikelets in rice (Oryza sativa L.). But the intrinsic factors responsible for the variations between the two types of spikelets are unclear. This

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service